  • Search Results
  • Electrochemical magnetoimmunosensor for the ultrasensitive determination of interleukin-6 in saliva and urine using poly-HRP streptavidin conjugates as labels for signal amplification.
Electrochemical magnetoimmunosensor for the ultrasensitive determination of interleukin-6 in saliva and urine using poly-HRP streptavidin conjugates as labels for signal amplification.
Analytical and bioanalytical chemistry (2014-08-02)
I Ojeda, M Moreno-Guzmán, A González-Cortés, P Yáñez-Sedeño, J M Pingarrón
ABSTRACT
A novel magnetoimmunosensor design for interleukin-6 (IL-6) which involved the covalent immobilization of anti-IL-6 antibodies onto carboxyl-functionalized magnetic microparticles and a sandwich-type immunoassay with signal amplification using poly-HRP-streptavidin conjugates is reported. All the variables concerning the preparation and the electroanalytical performance of the immunosensor were optimized. The use of poly-HRP-strept conjugates as enzymatic labels instead of conventional HRP-strept allowed enhanced signal-to-blank current ratios to be obtained. A linear calibration plot between the measured steady-state current and the log of IL-6 concentration was achieved in the 1.75 to 500 pg/mL range, which was not feasible when using HRP-strep as label. A limit of detection of 0.39 pg/mL IL-6 was obtained. The anti-IL-6-MB conjugates exhibited an excellent storage stability providing amperometric responses with no significant loss during at least 36 days. The magnetoimmunosensor showed also an excellent selectivity against potentially interfering substances. The immunosensor was used to determine IL-6 in urine samples spiked at three different concentration levels with clinical relevance. Moreover, IL-6 was measured in three different saliva samples corresponding to a periodontitis patient, a smoker volunteer, and a non-smoker volunteer. The obtained results were statistically in agreement with those provided by a commercial ELISA kit.
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Supelco
Ethanolamine, analytical standard
Sigma-Aldrich
Ethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Ethanolamine, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Aucubin, analytical standard
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
SAFC
Ethanolamine
Aucubin, primary reference standard
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
MES solution, BioUltra, for molecular biology, 0.5 M in H2O
Sigma-Aldrich
N-(3-Maleimidopropionyl)biocytin, ≥75% (HPLC)
Supelco
Hydroquinone, certified reference material, TraceCERT®