  • Search Results
  • Antiulcer activity of the ethanolic extract and ethyl acetate fraction of the leaves of Markhamia tomentosa in rats.
Antiulcer activity of the ethanolic extract and ethyl acetate fraction of the leaves of Markhamia tomentosa in rats.
Journal of ethnopharmacology (2014-09-23)
Margaret O Sofidiya, Foluso O Agunbiade, Neil A Koorbanally, Abimbola Sowemimo, Dapo Soesan, Titi Familusi
ABSTRACT
The leaves of Markhamia tomentosa (Benth.) K. Schum. Ex Engl. (Bignoniaceae) are used traditionally in the treatment of skin afflictions, sores, ulcers and inflammation. The aim of the study was to investigate the antiulcer activity of the crude ethanolic extract from the leaves of Markhamia tomentosa, determine the active fraction(s) of the extract and identify the chemical constituents in the active fraction by LC-MS. The antiulcer activity of the crude extract (50, 100 and 150mg/kg, p.o.) was evaluated in ethanol and indomethacin-induced models while the solvent fractions (150mg/kg) were screened using ethanol-induced gastric lesions in rats. Furthermore, anti-ulcer activity of the active fraction (50, 100 and 150mg/kg, p.o.) was performed using indomethacin and pylorus ligation models. Parameters such as gastric volume, pH and acidity were determined in the pylorus ligation model. LC-ESI-MS analysis was used to identify the components in the active fraction. The extract at the dose of 50, 100 and 150mg/kg caused a significant (p<0.05) dose-dependent inhibition of ulcer in the ethanol and indomethacin-induced ulcer models, respectively. The ethyl acetate (EtOAc) fraction showed the most potent antiulcer activity from all the fractions tested. This fraction produced 72% and 92% inhibition of indomethacin and pylorus-induced ulcer at a dose of 150mg/kg respectively. Acteoside, luteolin, luteolin-7-rutinoside, Luteolin-3',7-di-O-glucoside, carnosol, dilapachone, tormentic acid, oxo-pomolic acid and ajugol were detected in the EtOAc fraction. Our data provide a rational base for the folkloric use of Markhamia tormentosa in the treatment of ulcers.
Product Description
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, Preparateur, ≥99.9% (GC), Customized plastic drum
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, Preparateur, ≥99.9% (GC), One-time steel-plastic (SP) drum
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material