  • Search Results
  • Enantiomers separation by nano-liquid chromatography: use of a novel sub-2 μm vancomycin silica hydride stationary phase.
Enantiomers separation by nano-liquid chromatography: use of a novel sub-2 μm vancomycin silica hydride stationary phase.
Journal of chromatography. A (2015-01-24)
Silvia Rocchi, Anna Rocco, Joseph J Pesek, Maria T Matyska, Donatella Capitani, Salvatore Fanali
ABSTRACT
A novel sub-2 μm chiral stationary phase (CSP) was prepared immobilizing vancomycin onto 1.8 μm diol hydride-based silica particles. The CSP was packed into fused silica capillaries of 75 μm i.d. with a length of 11 cm and evaluated by means of nano-liquid chromatography (nano-LC) using model compounds of both pharmaceutical and environmental interest (some non-steroidal anti-inflammatory drugs, β-blockers and herbicides). The study of the effect of the linear velocity of the mobile phase on chromatographic efficiency showed good enantioresolutions up to a value of 5.11 at the optimal linear velocity with efficiencies in terms of number of plates per meter in the range 51,650-68,330. The results were compared with the ones obtained employing 5 μm vancomycin modified diol-silica particles packed in capillaries of the same i.d. For the acidic analytes the sub-2 μm CSP showed better performances, the baseline chiral separation of several studied compounds occurred in an analysis time of less than 3 min. Column-to-column packing reproducibility (n=3) expressed as relative standard deviation was in the range 2.2-5.8% and 0.5-7.7% for retention times and peak areas, respectively.
Sigma-Aldrich
Ketoprofen, ≥98% (TLC)
Sigma-Aldrich
Ketoprofen, meets USP testing specifications
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Supelco
Dichlorprop, PESTANAL®, analytical standard
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Supelco
Naproxen, VETRANAL®, analytical standard
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic, 99.998% trace metals basis
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Supelco
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Naproxen, meets USP testing specifications
Ketoprofen, European Pharmacopoeia (EP) Reference Standard
Supelco
Ketoprofen, VETRANAL®, analytical standard
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%