  • Search Results
  • Fine structure of the copulatory apparatus of the tapeworm Tetrabothrius erostris (Cestoda: Tetrabothriidea).
Fine structure of the copulatory apparatus of the tapeworm Tetrabothrius erostris (Cestoda: Tetrabothriidea).
Parasitology research (2015-04-10)
Janetta V Korneva, Malcolm K Jones, Vadim V Kuklin
ABSTRACT
The organization and fine structure of the complex copulatory apparatus of Tetrabothrius erostris (Tetrabothriidea) is investigated by light and transmission electron microscopy. A diversity of microstructures was found on the surface of genital ducts. The apical surfaces of male gonadoducts possess tubular and blade-like microtriches that have specific structure in each section of the duct. The apical part of the tubular microtriches contains numerous constrictions in the proximal section of the sperm duct; blade-like microtriches of cirrus possess longitudinal striation in the apical part, and their basal part is reinforced with electron-dense strands. Two types of microtriches occur on the surface of cirrus, and their presence may be considered as systematic features. Prostate glands containing granules of medium electron density (up to 130 nm diameter) are localized in the cirrus sac. The genital atrium contains numerous non-ciliated receptors. Paramyosin-like fibers (up to 200 nm) were found in the muscle fibers surrounding the male atrium canal. Microtriches on the surface of the distal region of the male atrial canal are covered by a glycocalyx. Electron-dense, membrane-like structures (up to 40 nm) lie under the apical membrane of the genital atrium and vagina. These structures do not form a continuous layer; its edges turn down and sink into the apical invaginations of epithelium. Hypotheses on the possible ways of copulation in T. erostris based on the observed ultrastructure are discussed.
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Osmium tetroxide solution, 4 wt. % in H2O
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Potassium hydroxide, anhydrous, ≥99.95% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Osmium tetroxide solution, 2.5 wt. % in tert-butanol
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol