  • Search Results
  • Hepatic microsomal UDP-glucuronosyltransferase (UGT) activities in the microminipig.
Hepatic microsomal UDP-glucuronosyltransferase (UGT) activities in the microminipig.
Biopharmaceutics & drug disposition (2014-04-23)
Eriko Higashi, Akihiro Ando, Shunsuke Iwano, Norie Murayama, Hiroshi Yamazaki, Yohei Miyamoto
ABSTRACT
The microminipig, a small minipig, was bred as a novel experimental animal for nonclinical pharmacology/toxicology studies by Fuji Micra Inc. (Shizuoka, Japan). Species differences in drug metabolism between humans and the microminipig need to be elucidated in more detail in order to discuss the results of nonclinical studies. Glucuronidation catalysed by UDP-glucuronosyltransferase (UGT) is an important pathway in the metabolism of a wide variety of compounds. The aim of the present study was to identify the characteristics of hepatic UGT activity in the microminipig and compare them with those in humans and other experimental animals. This study examined in vitro UGT activities using liver microsomes from microminipigs (8 months old and 1 day old), humans, mice, rats, dogs, monkeys and minipigs. The glucuronides of estradiol, imipramine, serotonin, propofol, 3'-azido-3'-deoxythymidine (AZT) and morphine, selective substrates of human UGT1A1, 1A4, 1A6, 1A9 and 2B7 (AZT and morphine), respectively, were measured using LC-MS/MS. Estradiol-3-glucuronidation activity was higher in the microminipig than in humans and the other animals. High levels of estradiol-3-glucuronidation were observed in the microsomes of 1-day-old microminipigs. Imipramine-N-glucuronidation, a distinctive conjugation by human UGT1A4, was catalysed by microminipig liver microsomes, but not by dog liver microsomes. Although AZT-glucuronidation activity was low in the microminipig compared with humans, morphine-3-glucuronidation activity in the microminipig was higher than that in humans. The UGT activities in the microminipig were similar to those in the minipig. The results of the present study have provided useful information for selecting an appropriate animal model for nonclinical studies.
Sigma-Aldrich
Serotonin hydrochloride, powder
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
Imipramine hydrochloride, ≥99% (TLC)
Sigma-Aldrich
2,6-Diisopropylphenol, 97%
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
3′-Azido-3′-deoxythymidine, ≥98% (HPLC)
Sigma-Aldrich
Uridine, BioUltra, ≥99%
Supelco
Zidovudine, Pharmaceutical Secondary Standard; Certified Reference Material
Propofol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Imipramine hydrochloride, BioXtra, ≥99% (TLC)
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Supelco
2,6-Diisopropylphenol, analytical standard
USP
Imipramine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Imipramine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Propofol for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Zidovudine, United States Pharmacopeia (USP) Reference Standard
Zidovudine, European Pharmacopoeia (EP) Reference Standard
Imipramine for system suitability, European Pharmacopoeia (EP) Reference Standard