  • Search Results
  • Differential organ phenotypes after postnatal Igf1r gene conditional deletion induced by tamoxifen in UBC-CreERT2; Igf1r fl/fl double transgenic mice.
Differential organ phenotypes after postnatal Igf1r gene conditional deletion induced by tamoxifen in UBC-CreERT2; Igf1r fl/fl double transgenic mice.
Transgenic research (2014-09-23)
Icíar Paula López, Lourdes Rodriguez-de la Rosa, Rosete Sofia Pais, Sergio Piñeiro-Hermida, Raquel Torrens, Julio Contreras, Isabel Varela-Nieto, José García Pichel
ABSTRACT
Insulin-like growth factor type 1 receptor (IGF1R) is a ubiquitously expressed tyrosine kinase that regulates cell proliferation, differentiation and survival. It controls body growth and organ homeostasis, but with specific functions depending on developmental time and cell type. Human deficiency in IGF1R is involved in growth failure, microcephaly, mental retardation and deafness, and its overactivation is implicated in oncogenesis. Igf1r-deficient mice die at birth due to growth retardation and respiratory failure. Although multiple Igf1r tissue-specific mutant lines have been analyzed postnatally, using Igf1r-floxed (Igf1r (fl/fl) ) mice mated with diverse cell-type recombinase Cre-expressing transgenics, no mouse models for the study of generalized Igf1r deficiency in adults have been reported. To this end we generated UBC-CreERT2; Igf1r (fl/fl) transgenic mice with an inducible deletion of Igf1r activated by tamoxifen. Tamoxifen administration to 4 week-old prepuberal male mice delayed their growth, producing a distinct impact on organ size 4 weeks later. Whereas testes were smaller, spleen and heart showed an increased organ to body weight ratio. Mosaic Igf1r genomic deletions caused a significant reduction in Igf1r mRNA in all organs analyzed, resulting in diverse phenotypes. While kidneys, spleen and cochlea had unaltered gross morphology, testes revealed halted spermatogenesis, and liver and alveolar lung parenchyma showed increased cell proliferation rates without affecting apoptosis. We demonstrate that UBC-CreERT2 transgenic mice efficiently delete Igf1r upon postnatal tamoxifen treatment in multiple mouse organs, and corroborate that IGF1R function is highly dependent on cell, tissue and organ type.
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
SAFC
Glycine
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, 99%, FCC
Glycine, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycine, certified reference material, TraceCERT®
Sigma-Aldrich
Solketal methacrylate, 50 wt. % in dichloromethane, contains ~280 ppm 4-tert-butylcatechol as inhibitor
Sigma-Aldrich
Glycine, tested according to Ph. Eur.