  • Search Results
  • Use of a carboxylesterase inhibitor of phenylmethanesulfonyl fluoride to stabilize epothilone D in rat plasma for a validated UHPLC-MS/MS assay.
Use of a carboxylesterase inhibitor of phenylmethanesulfonyl fluoride to stabilize epothilone D in rat plasma for a validated UHPLC-MS/MS assay.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-08-26)
Long Yuan, Yunlin Fu, Duxi Zhang, Yuan-Qing Xia, Qianping Peng, Anne-Françoise Aubry, Mark E Arnold
ABSTRACT
A sensitive, accurate and rugged UHPLC-MS/MS method was developed and validated for the quantitation of Epothilone D (EpoD), a microtubule stabilizer in development for treatment of Alzeimer's disease, in rat plasma. The ester group in EpoD can be hydrolyzed by esterases in blood or plasma, which creates a stability concern for the bioanalysis of EpoD. Species differences in the stability of EpoD in plasma were observed. Carboxylesterases were identified as the likely esterases responsible for the hydrolysis of EpoD in plasma ex vivo, and the cause of the species different stability. Phenylmethanesulfonyl fluoride, a carboxylesterase inhibitor, was used to stabilize EpoD in rat blood during sample collection, processing, and storage. A systematic method screening and optimization strategy was used to improve the assay sensitivity and minimize potential bioanalytical risks. The stabilized plasma samples were extracted by liquid-liquid extraction. Chromatographic separation was achieved on an Acquity UPLC BEH Phenyl column with a gradient elution. EpoD and its stable isotope labeled internal standards were detected by positive ion electrospray tandem mass spectrometry. The standard curve, which ranged from 0.100 to 100ng/mL was fitted to a 1/x(2) weighted linear regression model. The intra-assay precision was within ±3.6% CV and inter-assay precision was within ±4.2% CV. The assay accuracy was within ±8.3% of the nominal values. Assay recovery of EpoD was high (∼90%) and matrix effect was minimal (1.02-1.05). EpoD was stable in stabilized rat plasma for at least 30h at room temperature, 180 days at -20°C, and following three freeze-thaw cycles. The validated method was successfully applied to sample analysis in toxicology studies.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
1-Chlorobutane, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
1-Chlorobutane, anhydrous, 99.5%
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
1-Chlorobutane, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O