  • Search Results
  • Elevated plasma sphingomyelin (d18:1/22:0) is closely related to hepatic steatosis in patients with chronic hepatitis C virus infection.
Elevated plasma sphingomyelin (d18:1/22:0) is closely related to hepatic steatosis in patients with chronic hepatitis C virus infection.
European journal of clinical microbiology & infectious diseases : official publication of the European Society of Clinical Microbiology (2014-05-09)
J-F Li, F Qu, S-J Zheng, H-L Wu, M Liu, S Liu, Y Ren, F Ren, Y Chen, Z-P Duan, J-L Zhang
ABSTRACT
Hepatic steatosis affects disease progression in patients with chronic hepatitis C virus (HCV) infection. We investigated the plasma sphingolipid profile in patients with chronic hepatitis C (CHC) and whether there was an association between HCV-related steatosis and plasma sphingolipids. We used high-performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS) to analyze plasma sphingolipids in 120 interferon-naïve, non-diabetic, and non-obese CHC patients. Hepatic steatosis was defined as ≥5 % hepatocytes with fat based on histopathological analysis. Blood biochemical indicators and HCV load and genotype were also determined. Thirty-six (30.0 %) of 120 patients presented with hepatic steatosis Grades 1-3. Forty-four plasma sphingolipids were detected. Plasma sphingomyelin (SM) (d18:1/22:0) and ceramide (Cer) (d18:1/24:0)-1-P correlated with steatosis grade (r = 0.22, p = 0.015; r = -0.23, p = 0.012, respectively). SM (d18:1/22:0) [odds ratio (OR) = 1.12] and Cer (d18:1/24:0)-1-P (OR = 0.88) were independent factors for the presence of hepatic steatosis in CHC patients. The area under the curve (AUC) of SM (d18:1/22:0) and Cer (d18:1/24:0)-1-P was 0.637 and 0.638, respectively, to identify the presence of steatosis. Further analysis for genotype 2 CHC showed that only SM (d18:1/22:0) was independently linked to steatosis (OR = 1.21). The AUC of SM (d18:1/22:0) to identify hepatic steatosis in genotype 2 CHC was 0.726. Its sensitivity and negative predictive value reached 0.813 and 0.886, respectively. This study suggested that altered plasma SM (d18:1/22:0) was closely related to hepatic steatosis in chronic HCV infection, especially with genotype 2. Experimental studies are needed to determine further the underlying mechanisms responsible for these associations.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)