  • Search Results
  • Intestinal absorption and cell transforming potential of PhIP-M1, a bacterial metabolite of the heterocyclic aromatic amine 2-amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP).
Intestinal absorption and cell transforming potential of PhIP-M1, a bacterial metabolite of the heterocyclic aromatic amine 2-amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP).
Toxicology letters (2015-02-25)
Petra Nicken, Ina Willenberg, Anne von Keutz, Leonie von Elsner, Gerd Hamscher, Lynn Vanhaecke, Bernd Schröder, Gerhard Breves, Nils Helge Schebb, Pablo Steinberg
ABSTRACT
Previous studies have shown that in the rat, the colon carcinogen 2-amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP) is only absorbed to a limited extent in the small intestines and that a major fraction of unmetabolised PhIP reaches the colon. Moreover, PhIP is extensively metabolised when incubated with human stool samples to a major derivative, 7-hydroxy-5-methyl-3-phenyl-6,7,8,9-tetrahydropyrido [3',2':4,5]imidazo[1,2-a]pyrimidin-5-ium chloride (PhIP-M1). In the present study, the uptake and transport of PhIP-M1 in Ussing chamber experiments, its cytotoxicity in the different segments of the Fischer 344 rat gut and its transforming potential in the BALB/c 3T3 cell transformation assay were analysed. At the most, 10-20% of the PhIP-M1 amount added to the mucosal compartment of the Ussing chambers per segment were absorbed within 90min. Therefore, the amount of PhIP-M1 detected in the tissues as well as in the serosal compartment of the Ussing chambers was extremely low. Moreover, human-relevant concentrations of PhIP-M1 were not cytotoxic and did not induce the malignant transformation of BALB/c 3T3 cells. In conclusion, even if one would assume that 100% of the daily amount of PhIP ingested by a human being is converted into PhIP-M1 in the colon, this concentration most probably would not lead to cytotoxicity and/or carcinogenicity in the colorectal mucosa.
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
D-Mannitol, ≥98%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Magnesium chloride, powder, <200 μm