  • Search Results
  • Mycobacterium tuberculosis dysregulates MMP/TIMP balance to drive rapid cavitation and unrestrained bacterial proliferation.
Mycobacterium tuberculosis dysregulates MMP/TIMP balance to drive rapid cavitation and unrestrained bacterial proliferation.
The Journal of pathology (2014-09-05)
André Kübler, Brian Luna, Christer Larsson, Nicole C Ammerman, Bruno B Andrade, Marlene Orandle, Kevin W Bock, Ziyue Xu, Ulas Bagci, Daniel J Mollura, John Marshall, Jay Burns, Kathryn Winglee, Bintou Ahmadou Ahidjo, Laurene S Cheung, Mariah Klunk, Sanjay K Jain, Nathella Pavan Kumar, Subash Babu, Alan Sher, Jon S Friedland, Paul T G Elkington, William R Bishai
ABSTRACT
Active tuberculosis (TB) often presents with advanced pulmonary disease, including irreversible lung damage and cavities. Cavitary pathology contributes to antibiotic failure, transmission, morbidity and mortality. Matrix metalloproteinases (MMPs), in particular MMP-1, are implicated in TB pathogenesis. We explored the mechanisms relating MMP/TIMP imbalance to cavity formation in a modified rabbit model of cavitary TB. Our model resulted in consistent progression of consolidation to human-like cavities (100% by day 28), with resultant bacillary burdens (>10(7) CFU/g) far greater than those found in matched granulomatous tissue (10(5) CFU/g). Using a novel, breath-hold computed tomography (CT) scanning and image analysis protocol, we showed that cavities developed rapidly from areas of densely consolidated tissue. Radiological change correlated with a decrease in functional lung tissue, as estimated by changes in lung density during controlled pulmonary expansion (R(2)  = 0.6356, p < 0.0001). We demonstrated that the expression of interstitial collagenase (MMP-1) was specifically greater in cavitary compared to granulomatous lesions (p < 0.01), and that TIMP-3 significantly decreased at the cavity surface. Our findings demonstrated that an MMP-1/TIMP imbalance is associated with the progression of consolidated regions to cavities containing very high bacterial burdens. Our model provided mechanistic insight, correlating with human disease at the pathological, microbiological and molecular levels. It also provided a strategy to investigate therapeutics in the context of complex TB pathology. We used these findings to predict a MMP/TIMP balance in active TB and confirmed this in human plasma, revealing the potential of MMP/TIMP levels as key components of a diagnostic matrix aimed at distinguishing active from latent TB (PPV = 92.9%, 95% CI 66.1-99.8%, NPV = 85.6%; 95% CI 77.0-91.9%).
Product Description
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of Ph. Eur., BP, 35 wt. %, contains 10% methanol as stabilizer
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O