  • Search Results
  • Engineering the oxygen sensing regulation results in an enhanced recombinant human hemoglobin production by Saccharomyces cerevisiae.
Engineering the oxygen sensing regulation results in an enhanced recombinant human hemoglobin production by Saccharomyces cerevisiae.
Biotechnology and bioengineering (2014-08-02)
José L Martínez, Lifang Liu, Dina Petranovic, Jens Nielsen
ABSTRACT
Efficient production of appropriate oxygen carriers for transfusions (blood substitutes or artificial blood) has been pursued for many decades, and to date several strategies have been used, from synthetic polymers to cell-free hemoglobin carriers. The recent advances in the field of metabolic engineering also allowed the generation of different genetically modified organisms for the production of recombinant human hemoglobin. Several studies have showed very promising results using the bacterium Escherichia coli as a production platform, reporting hemoglobin titers above 5% of the total cell protein content. However, there are still certain limitations regarding the protein stability and functionality of the recombinant hemoglobin produced in bacterial systems. In order to overcome these limitations, yeast systems have been proposed as the eukaryal alternative. We recently reported the generation of a set of plasmids to produce functional human hemoglobin in Saccharomyces cerevisiae, with final titers of active hemoglobin exceeding 4% of the total cell protein. In this study, we propose a strategy for further engineering S. cerevisiae by altering the oxygen sensing pathway by deleting the transcription factor HAP1, which resulted in an increase of the final recombinant active hemoglobin titer exceeding 7% of the total cellular protein.
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Potassium hydroxide, anhydrous, ≥99.95% trace metals basis
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E422, anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous