Point-of-care fluorescence immunoassay for cardiac panel biomarkers.
Journal of clinical laboratory analysis (2014-03-22)
Tae Kyum Kim, Sang Wook Oh, Soon Cheol Hong, Young Joon Mok, Eui Yul Choi
ABSTRACT
Myoglobin, creatine kinase-MB isoenzyme (CK-MB), and cardiac troponin I (cTnI) are cardiac biomarkers that are widely used to assist in the early and late diagnoses of acute myocardial infarction (AMI). Here, we present a clinically applicable fluorescence (FL) immunoassay for cardiac biomarkers. Whole blood was mixed with FL-labeled detector Ab (antibody) and then loaded onto a capture Ab-immobilized strip in a test cartridge. The FL intensities at test and control line on the strip were obtained and converted in a laser FL scanner to determine the concentration of biomarker. The analytical performance of immunoassay system was evaluated by linearity and imprecision tests. The comparability of the FL immunoassay method was examined with a reference method. FL intensities and the levels of myoglobin, CK-MB, and cTnI displayed good linearity and high correlations (r = 0.999, 0.998, and 0.989, respectively). The coefficient of variations (CVs) of imprecision for all cardiac biomarkers were less than 8% in both intra- and interassays. When the results from the developed method and bioMerieux VIDAS assay were analyzed by Bland-Altman plot and Passing-Bablok plot, the two assay methods were in good agreement. The FL immunoassay system can provide a platform for point-of-care testing (POCT), and it is an easy, fast, and reliable method for the quantification of cardiac biomarkers.
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C