  • Search Results
  • Statistical significance of hair analysis of clenbuterol to discriminate therapeutic use from contamination.
Statistical significance of hair analysis of clenbuterol to discriminate therapeutic use from contamination.
Drug testing and analysis (2014-11-13)
Aniko Krumbholz, Patricia Anielski, Lena Gfrerer, Matthias Graw, Hans Geyer, Wilhelm Schänzer, Jiri Dvorak, Detlef Thieme
ABSTRACT
Clenbuterol is a well-established β2-agonist, which is prohibited in sports and strictly regulated for use in the livestock industry. During the last few years clenbuterol-positive results in doping controls and in samples from residents or travellers from a high-risk country were suspected to be related the illegal use of clenbuterol for fattening. A sensitive liquid chromatography-tandem mass spectrometry (LC-MS/MS) method was developed to detect low clenbuterol residues in hair with a detection limit of 0.02 pg/mg. A sub-therapeutic application study and a field study with volunteers, who have a high risk of contamination, were performed. For the application study, a total dosage of 30 µg clenbuterol was applied to 20 healthy volunteers on 5 subsequent days. One month after the beginning of the application, clenbuterol was detected in the proximal hair segment (0-1 cm) in concentrations between 0.43 and 4.76 pg/mg. For the second part, samples of 66 Mexican soccer players were analyzed. In 89% of these volunteers, clenbuterol was detectable in their hair at concentrations between 0.02 and 1.90 pg/mg. A comparison of both parts showed no statistical difference between sub-therapeutic application and contamination. In contrast, discrimination to a typical abuse of clenbuterol is apparently possible. Due to these findings results of real doping control samples can be evaluated.
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Potassium hydroxide, anhydrous, ≥99.95% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane