UGT1A1*28 polymorphism influences glucuronidation of bazedoxifene.
Die Pharmazie (2015-05-23)
T Trdan Lušin, A Mrhar, J Trontelj
ABSTRACT
Bazedoxifene is used for the prevention and treatment of osteoporosis. After peroral application, bazedoxifene is metabolized by UDP-glucuronosyltransferases (UGTs) to bazedoxifene-4'-glucuronide (M4) and bazedoxifene-5-glucuronide (M5). It has already been shown that a relatively common UGT1A1*28 polymorphism can considerably affect raloxifene pharmacokinetics and pharmacodynamics. As pharmacokinetics of bazedoxifene and raloxifene are very similar, the influence of UGT1A1*28 polymorphism on metabolism of bazedoxifene was investigated by genotyped microsomes. Our results indicate an influence of UGT1A1*28 allele on the formation clearance of both bazedoxifene metabolites. The decreased metabolic clearance was most pronounced in microsomes from polymorphic homozygote (*28/*28) where a 7 to 10-fold lower metabolic clearance was observed for both metabolites compared to other genotypes. In conclusion, the significant UGT1A1*28 genotype effect on bazedoxifene intrinsic metabolic clearance indicates that this subject is worth further exploration in vivo and provides valuable information research in this field.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
D-Glucuronic acid, ≥98% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Haloperidol, powder
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Uridine, BioUltra, ≥99%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Haloperidol, European Pharmacopoeia (EP) Reference Standard
USP
Haloperidol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Haloperidol for peak identification, European Pharmacopoeia (EP) Reference Standard
Haloperidol for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)