  • Search Results
  • Danzhi Xiaoyao San ameliorates depressive-like behavior by shifting toward serotonin via the downregulation of hippocampal indoleamine 2,3-dioxygenase.
Danzhi Xiaoyao San ameliorates depressive-like behavior by shifting toward serotonin via the downregulation of hippocampal indoleamine 2,3-dioxygenase.
Journal of ethnopharmacology (2014-12-02)
Xiaoxia Zhu, Linlin Jing, Chun Chen, Meng Shao, Qin Fan, Jianxin Diao, Yuanliang Liu, Zhiping Lv, Xuegang Sun
ABSTRACT
Danzhi Xiaoyao San (DXS) is a canonical Chinese medicine formula from Principles of Internal Medicine, which was written during the Ming dynasty. This formula is approved and commercialized for use in the prevention and treatment of affective disorders. This study is aimed to investigate the hypothesis that DXS treats depressive-like behavior by shifting the balance of the kynurenine (Kyn)/serotonin (5-HT) pathway toward the 5-HT pathway through the downregulation of hippocampal indoleamine 2,3-dioxygenase (IDO). Chemical fingerprints of gardenoside, paeoniflorin, ferulic acid, paeonol, and ligustilide in standard extraction were used as the material bases of DXS. Rats with depressive-like behavior induced by chronic unpredictable mild stress (CUMS) were randomly divided into four groups, namely the control, model, DXS, and fluoxetine groups. Cytokines, IDO, and tryptophan (Trp) catabolites were analyzed by enzyme-linked immunosorbent assay, western blot, and liquid chromatography-electrospray ionization tandem mass spectrometry, respectively. DXS significantly increased crossing grid numbers, sucrose consumption, and body weight. This treatment significantly decreased the serum levels of tumor necrosis factor-α and interleukin 6 (IL-6). However, DXS elicited no significant effects on IL-1β, IL-2, and interferon γ. DXS downregulated the activity of IDO and subsequent production of Kyn in the hippocampus. This treatment upregulated the hippocampal contents of Trp and 5-HT but did not influence 5-HT turnover. DXS exhibited antidepressant-like effects on rats exposed to CUMS. DXS reduced IDO activity to shift the balance of the Kyn/5-HT pathway toward the 5-HT pathway.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Serotonin hydrochloride, powder
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Fluoxetine hydrochloride, solid
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Kynurenine, ≥98% (HPLC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)