  • Search Results
  • Cystic fibrosis sputum DNA has NETosis characteristics and neutrophil extracellular trap release is regulated by macrophage migration-inhibitory factor.
Cystic fibrosis sputum DNA has NETosis characteristics and neutrophil extracellular trap release is regulated by macrophage migration-inhibitory factor.
Journal of innate immunity (2014-05-28)
Markryan Dwyer, Qiang Shan, Samantha D'Ortona, Rie Maurer, Robert Mitchell, Hanne Olesen, Steffen Thiel, Johannes Huebner, Mihaela Gadjeva
ABSTRACT
Neutrophils are the main proinflammatory cell type in chronically infected lungs of cystic fibrosis (CF) patients; however, they fail to effectively clear the colonizing pathogens. Here, we investigated the molecular composition of non-mucoid and mucoid Pseudomonas aeruginosa-induced neutrophil extracellular traps (NETs) in vitro and compared them to the DNA-protein complexes present in the CF sputum. The protein composition of P. aeruginosa-induced NET fragments revealed that irrespective of the inducing stimuli, NET fragments were decorated with a conserved set of proteins. The DNA-protein complexes derived from CF sputum were consistent with NETosis and shared a similar protein signature, suggesting that the majority of the extracellular DNA was NET derived. The ability of polymorphonuclear leukocytes to produce NETs in response to P. aeruginosa was driven by macrophage migration-inhibitory factor (MIF) by promoting mitogen-activated protein kinase. Analysis of 132 CF patient samples revealed that elevated MIF protein levels correlated with poorer lung function. We suggest that targeting MIF by small molecular inhibitors might reduce the presence of extracellular DNA and serve as an adjunct to the use of antimicrobial drugs that could ultimately reduce bacterial fitness in the lungs during the later stages of CF disease.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of Ph. Eur., BP, 35 wt. %, contains 10% methanol as stabilizer
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
HEPES
Sigma-Aldrich
HEPES, Vetec, reagent grade, 99.5%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
SAFC
HEPES
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Lithium dodecyl sulfate, BioXtra, ≥98.5% (GC)
Sigma-Aldrich
Lithium dodecyl sulfate, BioReagent, for molecular biology, suitable for electrophoresis
Sigma-Aldrich
Lithium dodecyl sulfate, ≥98.5% (GC)
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sucrose, ≥99.5% (GC)