  • Search Results
  • Ferrocene-based guanidine derivatives: in vitro antimicrobial, DNA binding and docking supported urease inhibition studies.
Ferrocene-based guanidine derivatives: in vitro antimicrobial, DNA binding and docking supported urease inhibition studies.
European journal of medicinal chemistry (2014-08-12)
Rukhsana Gul, Muhammad Khawar Rauf, Amin Badshah, Syed Sikander Azam, Muhammad Nawaz Tahir, Azim Khan
ABSTRACT
Some novel ferrocenyl guanidines 1-8 were synthesized and characterized by different spectroscopic methods, elemental analysis and single crystal X-rays diffraction techniques. The crystallographic studies revealed that the existence of the strong non-bonding interactions facilitate these molecules to interact with biological macro-molecules like DNA that described to inherit good biological activities. The DNA interaction studies carried out by cyclic voltammetry (CV) and UV-visible spectroscopy are in close agreement with the binding constants (K) (0.79-5.4) × 10(5) (CV) and (0.72-5.1) × 10(5) (UV-vis). The shift in peak potential, current and absorption maxima of the studied ferrocenyl guanidines in the presence of DNA revealed that CV coupled with UV-vis spectroscopy could provide an opportune to characterize metal-based compounds-DNA interaction mechanism, a prerequisite for the design of new anticancer agents and understanding the molecular basis of their action. The compounds 1-8 have been screened for their antibacterial, antifungal and urease inhibition potency. A concurrent in silico study has also been applied on ferrocene moiety impregnated guanidines 1-8 to identify most active compounds having for inhibiting the activity of urease (pdb id 3LA4). Most of the compounds were found as potent inhibitors of urease and the compound 1 was found to be the most active with an IC50 of 16.83 ± 0.03 μM. The docking scores are in close agreement with the in vitro obtained IC50 values of inhibitors 1-8.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Ferrocene, 98%
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Diethyl ether, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Supelco
Melting point standard 121-123°C, analytical standard
Sigma-Aldrich
Sodium nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
4-Nitroaniline, ≥99%