  • Search Results
  • Nanoparticle formulation improves the anticonvulsant effect of clonazepam on the pentylenetetrazole-induced seizures: behavior and electroencephalogram.
Nanoparticle formulation improves the anticonvulsant effect of clonazepam on the pentylenetetrazole-induced seizures: behavior and electroencephalogram.
Journal of pharmaceutical sciences (2014-06-12)
Gerardo Leyva-Gómez, María Eva González-Trujano, Edith López-Ruiz, Pierre-Olivier Couraud, Babette Wekslerg, Ignacio Romero, Florence Miller, Florence Delie, Eric Allémann, David Quintanar-Guerrero
ABSTRACT
To document the efficacy of clonazepam (CLZ) either free as a solution or loaded in solid lipid nanoparticles (CLZ-SLN) or mixed micelles (CLZ-MM), the in vitro blood-brain barrier permeability of the formulations was determined. Behavior and/or electroencephalograms (EEGs) of rodents receiving treatments were also studied. The in vitro permeability of CLZ increased when associated with SLN, but decreased in the case of MM. The occurrence of the pentylenetetrazole (PTZ)-induced seizures in mice was significantly prevented by CLZ, even when exposed a lower dose of CLZ-SLN after administration by the oral route. The behavioral severity and EEGs showing the PTZ-induced paroxystic activity in rats diminished significantly in the presence of CLZ alone (0.3 mg/kg), and were almost totally prevented in the rats treated with CLZ-SLN (equivalent to 0.3 mg/kg). The frequency, duration, and spreading of the spikes-wave of rats treated with CLZ-SLN decreased significantly as compared with CLZ alone, CLZ-MM, or the vehicle. These results show an in vitro-in vivo correlation in the enhanced blood-brain barrier permeability of SLN formulation, and a contribution of MM to the carrier effect of drugs toward the bloodstream and brain, where this pharmaceutical formulation of CLZ-SLN improves the anticonvulsant effect of this benzodiazepine, thus offering additional advantages after oral administration.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
SAFC
HEPES