  • Search Results
  • Celastrol prevents circulatory failure via induction of heme oxygenase-1 and heat shock protein 70 in endotoxemic rats.
Celastrol prevents circulatory failure via induction of heme oxygenase-1 and heat shock protein 70 in endotoxemic rats.
Journal of ethnopharmacology (2015-01-13)
Yi-Li Wang, Kwok-Keung Lam, Pao-Yun Cheng, Yen-Mei Lee
ABSTRACT
Celastrol, a quinone methide extracted from the root of Tripterygium wilfordii Hook, possesses anti-oxidant and anti-inflammatory effects. Tripterygium wilfordii Hook is officially listed in the Chinese Pharmacopoeia and is used traditionally against rheumatoid arthritis, ankylosing spondylitis, and cancer. Furthermore, the circulatory protective effect of celastrol on an in vivo animal model of sepsis was investigated. Sepsis is a systemic inflammatory disorder that increases tissue oxidative stress and leads to multiple organ injury. We evaluated the beneficial effects of celastrol on multiple organ failure induced by lipopolysaccharide (LPS) in rats. Celastrol (0.5 and 1.0 mg/kg, i.v.) was administered to anaesthetized rats 2 h before and 30 min after LPS challenge (10 mg/kg, i.v.). Eight hours later, cardiac and aortic protein expressions related to inflammatory responses, superoxide anion production, and reduced glutathione (GSH) level were measured. Treatment with celastrol prevented circulatory failure (bradycardia and hypotension) 8h after LPS challenge. The plasma levels of ALT, LDH, TNF-α, and nitric oxide metabolites increased markedly during sepsis, which significantly reduced after celastrol treatments. Celastrol attenuated iNOS, TNF-α, NF-κB phospho-p65 expression, superoxide anion production, and caspase 3 activity in the cardiovascular system, all of which were markedly elevated after LPS challenge. Furthermore, celastrol induced HO-1 and HSP70 expressions increase in nuclear levels of Nrf2 and HSF-1, respectively, and increase cardiac GSH level 8h after LPS challenge. Anti-inflammatory and anti-oxidant effects of celastrol contribute to prevent circulatory failure in sepsis. Induction of HO-1 and HSP70 by celastrol participates in these beneficial effects.
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Supelco
Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl, saturated with silver chloride
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Sodium nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Potassium Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Supelco
Sodium nitrate, analytical standard
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Phenylephrine, European Pharmacopoeia (EP) Reference Standard
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%