  • Search Results
  • Structure of the N-terminal segment of human retinol dehydrogenase 11 and its preferential lipid binding using model membranes.
Structure of the N-terminal segment of human retinol dehydrogenase 11 and its preferential lipid binding using model membranes.
Biochimica et biophysica acta (2014-12-30)
Mustapha Lhor, Mario Méthot, Habib Horchani, Christian Salesse
ABSTRACT
Retinol dehydrogenase 11 (RDH11) has been postulated to be anchored to membranes by means of its N-terminal segment in retinal pigment epithelial (RPE) cells where it participates to the visual cycle. The analysis of the primary sequence of RDH11 revealed that its N-terminal hydrophobic segment could be involved in the anchoring of this enzyme to membranes. However, no information is yet available on the properties of this N-terminal segment to support this role. The secondary structure and membrane binding of two N-terminal peptides of RDH11 with different lengths have thus been investigated to provide this information. Online tools allowed predicting an α-helical secondary structure for both peptides. Infrared spectroscopy and circular dichroism have shown that the α-helix of the Long-peptide (35 amino acids) is longer and more rigid than that of the Short-peptide (25 amino acids) regardless of the type of solvent. Langmuir monolayers have been used as a model membrane to study lipid-peptide interactions. Values of maximum insertion pressure and synergy suggested a preferential binding of the Long-peptide to lipids with a phosphoethanolamine polar head group, which are abundant in the RPE. Furthermore, infrared spectroscopy in monolayers has shown that the α-helical structure of the Long-peptide is more stable in the presence of saturated phospholipids whereas the structure of the Short-peptide is mainly disordered. Altogether, the present data demonstrate that the α-helical hydrophobic core of the N-terminal segment of RDH11 displays properties typical of transmembrane domains, in agreement with its postulated role in the membrane anchoring of this protein.
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Supelco
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, purum, ≥99.0% (GC)
Sigma-Aldrich
Argon, ≥99.998%
USP
Butylated hydroxytoluene, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
O-Phospho-L-serine
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine, ≥99.0% (10 mg phospholipid per ml CHCl3, TLC)
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine, ≥97.0% (TLC)
Sigma-Aldrich
L-α-Phosphatidylethanolamine, dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder
Sigma-Aldrich
1,2-Distearoyl-sn-glycero-3-phosphoethanolamine, ≥99%
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine, ≥97%
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, tested according to Ph. Eur.
Sigma-Aldrich
Argon-40Ar, 99.95 atom %