  • Search Results
  • Reduced graphene oxide nanosheets coated with an anti-angiogenic anticancer low-molecular-weight heparin derivative for delivery of anticancer drugs.
Reduced graphene oxide nanosheets coated with an anti-angiogenic anticancer low-molecular-weight heparin derivative for delivery of anticancer drugs.
Journal of controlled release : official journal of the Controlled Release Society (2014-06-29)
Gayong Shim, Ji-Young Kim, Jeonghoon Han, Seung Woo Chung, Soondong Lee, Youngro Byun, Yu-Kyoung Oh
ABSTRACT
Here, we report reduced graphene oxide (rGO) nanosheets coated with an anti-angiogenic anticancer taurocholate derivative of low-molecular-weight heparin (LHT7) as a tumor-targeting nanodelivery platform for anticancer drugs. Surface coating of LHT7 onto rGO was confirmed using fluorescein isothiocyanate-labeled LHT7, monitored as fluorescence quenching due to associated rGO. Unlike plain rGO, LHT7-coated rGO (LHT-rGO) nanosheets maintained a stable dispersion under physiological conditions for at least 24h. Moreover, LHT-rGO provided greater loading capacity for doxorubicin (Dox) compared with uncoated rGO nanosheets. Following intravenous administration into KB tumor-bearing mice, in vivo tumor accumulation of LHT-rGO/Dox was 7-fold higher than that of rGO/Dox 24h post dosing. In tumor tissues, LHT-rGO/Dox was shown to localize not to the tumor vasculature, but rather to tumor cells. Intravenously administered LHT-rGO/Dox showed the greatest anti-tumor effect in KB-bearing mice, reducing tumor volume by 92.5%±3.1% compared to the untreated group 25days after tumor inoculation. TUNEL assays revealed that the population of apoptotic cells was highest in the group treated with LHT-rGO/Dox. Taken together, our results demonstrate that LHT-rGO nanosheets confer improved dispersion stability, tumor distribution and in vivo antitumor effects, and may be further developed as a potential active nanoplatform of various anticancer drugs.
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Ammonia solution, 2.0 M in methanol
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Potassium permanganate, purum p.a., ≥99.0% (RT), fine crystals
Sigma-Aldrich
Potassium permanganate, meets USP testing specifications