  • Search Results
  • Enhanced reproducibility of the human gel-based tear proteome maps in the presence of di-(2-hydroxyethyl) disulfide.
Enhanced reproducibility of the human gel-based tear proteome maps in the presence of di-(2-hydroxyethyl) disulfide.
Biotechnology and applied biochemistry (2014-03-01)
Neda Saraygord-Afshari, Hossein Naderi-Manesh, Mostafa Naderi
ABSTRACT
Patterns obtained in two-dimensional gel electrophoresis (2-DE) in the previously published articles suggest a varying number of proteins. To seek the cause of this variation, we investigated the effect of reduction power on the overall tear proteome maps. To this end, the buffers of two reducing agents, dithiothreitol (DTT) at nine different concentrations and di-(2-hydroxyethyl) disulfide (HED), were examined. The assay showed that HED clearly improved 2-DE resolution, increased the number of detectable protein spots, and offered well-resolved chain regions in comparison with those treated with DTT. Furthermore, this study introduced increasing the reduction power as a remedy to increase the reproducibility of two-dimensional human tear proteome maps. In addition, the results of our assessment showed that improved reduction efficiency was accompanied by increased procedure reproducibility from 42% to 89%.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Bromophenol Blue
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri