  • Search Results
  • Chemical composition, antimicrobial activity against Staphylococcus aureus and a pro-apoptotic effect in SGC-7901 of the essential oil from Toona sinensis (A. Juss.) Roem. leaves.
Chemical composition, antimicrobial activity against Staphylococcus aureus and a pro-apoptotic effect in SGC-7901 of the essential oil from Toona sinensis (A. Juss.) Roem. leaves.
Journal of ethnopharmacology (2014-04-15)
Jian-Guo Wu, Wei Peng, Jun Yi, Yan-Bin Wu, Ti-Qiang Chen, Ka-Hing Wong, Jin-Zhong Wu
ABSTRACT
Leaves of Toona sinensis (A. Juss.) Roem. (TSL), a popular vegetable in China, have anti-inflammatory, antidoting, and worm-killing effects and are used in folk medicine for the treatment of enteritis, dysentery, carbuncles, boils, and especially abdominal tumors. Our aim was to investigate the in vitro antimicrobial activity against Staphylococcus aureus and anticancer property of the essential oil from TSL (TSL-EO), especially the pro-apoptotic effect in SGC-7901. TSL-EO obtained by hydrodistillation was analyzed by GC/MS and was tested in vitro against twenty clinically isolated strains of Staphylococcus aureus (SA 1-20), which were either methicillin-sensitive Staphylococcus aureus (MSSA) or methicillin-resistant Staphylococcus aureus (MRSA) and two standard strains viz. ATCC 25923 and ATCC 43300. The anticancer activity of TSL-EO was evaluated in vitro against HepG2, SGC7901, and HT29 through MTT assay. Moreover, the apoptosis-inducing activity of TSL-EO in SGC7901 cells was determined by Hoechst 33324 staining and flow cytometry methods. Also, the apoptosis-related proteins viz. Bax, Bcl-2 and caspase-3 were detected by western-blotting. GC-MS analysis showed that TSL-EO contained a high amount of sesquiterpenes (84.64%), including copaene (8.27%), β-caryophyllene (10.16%), caryophyllene (13.18%) and β-eudesmene (5.06%). TSL-EO inhibited the growth of both MSSA and MRSA, with the lowest MIC values of 0.125 and 1mg/ml, respectively. Treatment with TSL-EO for 24h could significantly suppress the viability of three different cancer cell lines (P<0.05). Furthermore, the apoptosis-inducing activity of TSL-EO in SGC7901 cells increased in a dose-dependent manner, potentially resulting from the up-regulated expression of Bax, caspase-3 and down-regulated expression of Bcl-2. TSL-EO possessed antibacterial activity against Staphylococcus aureus and significant cytotoxicity against cancer cells and particularly prominent pro-apoptotic activity in SGC7901 cells. These bioactivities were probably due to the high content of sesquiterpenes. Our results suggested that TSL-EO possessed potential health benefits and could serve as a promising natural food addictive.
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Supelco
Hexane, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous