  • Search Results
  • Extracellular HCV-core protein induces an immature regulatory phenotype in NK cells: implications for outcome of acute infection.
Extracellular HCV-core protein induces an immature regulatory phenotype in NK cells: implications for outcome of acute infection.
PloS one (2014-07-31)
Lucy Golden-Mason, Young S Hahn, Michael Strong, Linling Cheng, Hugo R Rosen
ABSTRACT
Hepatitis C viral (HCV) proteins, including core, demonstrate immuno-modulatory properties; however, the effect of extracellular core on natural killer (NK) cells has not previously been investigated. To characterise NKs in acute HCV infection over time, and, to examine the effect of exogenous HCV-core protein on NK cell phenotype and function. Acute HCV patients (n = 22), including 10 subjects who spontaneously recovered, were prospectively studied. Flow-cytometry was used to measure natural cytotoxicity and to phenotype NKs directly ex vivo and after culture with HCV-core protein. Microarray analysis was used to identify pathways involved in the NK cell response to exogenous HCV-core. Direct ex vivo analysis demonstrated an increased frequency of immature/regulatory CD56bright NKs early in acute HCV infection per se which normalized with viral clearance. Natural cytotoxicity was reduced and did not recover after viral clearance. There was a statistically significant correlation between the frequency of CD56bright NKs and circulating serum levels of HCV core protein. In vitro culture of purified CD56bright NK cells with HCV-core protein in the presence of IL-15 maintained a significant proportion of NKs in the CD56bright state. The in vitro effect of core closely correlates with NK characteristics measured directly ex vivo in acute HCV infection. Pathway analysis suggests that HCV-core protein attenuates NK interferon type I responses. Our data suggest that HCV-core protein alters NK cell maturation and may influence the outcome of acute infection.
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%