  • Search Results
  • Identification and functional traits of lactic acid bacteria isolated from Ciauscolo salami produced in Central Italy.
Identification and functional traits of lactic acid bacteria isolated from Ciauscolo salami produced in Central Italy.
Meat science (2014-08-05)
Sara Federici, Floriana Ciarrocchi, Raffaella Campana, Eleonora Ciandrini, Giuliana Blasi, Wally Baffone
ABSTRACT
Lactic acid bacteria (LAB) from Ciauscolo salami produced in Marche Region of Central Italy, and LAB strains belonging to our laboratory collection were examined for their capability to survive at low pH and bile, to adhere to Caco-2 cells, and for antibiotic resistance. LAB from Ciauscolo were identified by ARDRA and RAPD-PCR. Our study showed that all LAB strains had good adaptation to gastric juice and moderate tolerance to bile. The adhesiveness was variable among strains but significantly lower in LAB from food. Antibiotic resistance was broadly spread among food strains, with level of resistance exceeding 15% for all the antibiotics tested. The resistance determinants erm(B) and tet(M) were found in nine strains of food origin (21.4%) while tet(L) in one strain of our collection (5%). Our work suggests that fermented foods are valuable sources of bacterial strains with functional traits of intestinal lactobacilli. These bacteria may be further studied for their use in probiotic applications.
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Sigma-Aldrich
Lactic acid, 85%, FCC
Sigma-Aldrich
DL-Lactic acid, ~90% (T)
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Sigma-Aldrich
Lactic acid, natural, ≥85%
Supelco
Lactic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
USP
Lactic acid, United States Pharmacopeia (USP) Reference Standard