  • Search Results
  • Leishmanicidal activity of polysaccharides and their oxovanadium(IV/V) complexes.
Leishmanicidal activity of polysaccharides and their oxovanadium(IV/V) complexes.
European journal of medicinal chemistry (2014-12-17)
Alex Evangelista do Amaral, Carmen Lúcia Oliveira Petkowicz, Ana Lucia Ramalho Mercê, Marcelo Iacomini, Glaucia Regina Martinez, Maria Eliane Merlin Rocha, Silvia Maria Suter Correia Cadena, Guilhermina Rodrigues Noleto
ABSTRACT
The parasites of the genus Leishmania cause a range of leishmaniasis diseases, whose treatment is impaired due to intramacrophage parasites living in the mammalian host. Immunostimulation has been considered an important strategy to leishmaniasis treatment. The immunomodulatory effects of the polysaccharides arabinogalactan (ARAGAL), galactomannan (GMPOLY), and xyloglucan (XGJ), as well as their oxovanadium (IV/V) complexes (ARAGAL:VO, GMPOLY:VO, and XGJ:VO) were evaluated on peritoneal macrophages. At 25 μg/mL of GMPOLY:VO and of XGJ:VO, and 10 μg/mL of ARAGAL:VO, nitric oxide (NO) production by the macrophages was not altered compared with the control group. All polymers increased the production of interleukins 1 beta and 6 (IL-1β and IL-6), but the oxovanadium complexes were more potent activators of these mediators. ARAGAL:VO 10 μg/mL, GMPOLY:VO and XGJ:VO 25 μg/mL led to an increase of 562%, 1054%, and 523% for IL-1β, respectively. For IL-6 at the same concentration, the levels increased by 539% and 794% for ARAGAL:VO and GMPOLY:VO, respectively. Polysaccharides and their oxovanadium complexes exhibited important leishmanicidal effects on amastigotes of Leishmania (L.) amazonensis. The native and complexed polymers reduced the growth of promastigote-form Leishmania by ∼60%. This effect was reached at concentrations 12 times lower than that observed for Glucantime (300 μg/mL promoted an inhibition of ∼60%). The 50% inhibitory concentration (IC50) values for the complexes were determined. XGJ:VO showed the lowest IC50 value (6.2 μg/mL; 0.07 μg/mL of vanadium), which for ARAGAL:VO was 6.5 μg/mL (0.21 μg/mL of vanadium) and 7.3 μg/mL (0.06 μg/mL of vanadium) for GMPOLY:VO. The upregulation of IL-1β and IL-6 release and downregulation of NO production by macrophages and the important leishmanicidal effect are essential to stablish their potential use against this pathology.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sulfanilamide, ≥98%
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
SAFC
HEPES
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Supelco
Sulfanilamide melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥99% (calc. to the dried substance)
Sigma-Aldrich
1-(2-Hydroxyethyl)piperazine, 98%
Sigma-Aldrich
Potassium hydroxide, anhydrous, ≥99.95% trace metals basis
SAFC
HEPES
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Sigma-Aldrich
Potassium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets
USP
Sulfanilamide Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
USP
Sulfanilamide, United States Pharmacopeia (USP) Reference Standard