Merck
Proteomic analysis of exosomes derived from human lymphoma cells.
European journal of medical research (2015-01-30)
Ye Yao, Wei Wei, Jing Sun, Linjun Chen, Xiaohui Deng, Liyuan Ma, Siguo Hao
ABSTRACT
Exosomes secreted by tumor cells contain specific antigens that may have immunotherapeutic purposes. The aim of this study was to characterize the proteomic content of lymphoma cell-derived exosomes (LCEXs). In this study, exosomes derived from Raji cells (EXO(Raji)) were purified and proteins of EXO(Raji) were separated by one-dimensional sodium dodecyl sulfate polyacrylamide gel electrophoresis. Protein bands were identified by mass spectrometry. The protein components of EXO(Raji) were analyzed using shotgun technology, and the function proteins of EXO(Raji) were defined and described using the Gene Ontology (GO) database and Kyoto Encyclopedia of Genes and Genomes (KEGG) analysis. A total of 197 proteins were identified in EXO(Raji); 139 proteins were also identified in Raji cells, showing an overlap of 70.56% of the total proteins in EXO(Raji). Interestingly, the remaining 58 proteins were unique to EXO(Raji). The GO database and KEGG were used to define and describe the function of proteins. The data showed that some important proteins involved in antigen procession and presentation as well as cell migration and adhesion were also identified in EXO(Raji), such as MHC-I and II, HSC70, HSP90, and ICMA-1. LCEXs express a discrete set of proteins involved in antigen presentation and cell migration and adhesion, suggesting that LCEXs play an important role in the regulation of immunity and interaction between lymphoma cells and their microenvironment. LCEXs harbor most of the proteins of lymphoma cells and could be one of the sources of lymphoma-associated antigens for immunotherapeutic purposes.
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Deuterium oxide, "100%", 99.990 atom % D
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Deuterium oxide, standard, 99.98 atom %±0.01 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)