  • Search Results
  • Betanin attenuates paraquat-induced liver toxicity through a mitochondrial pathway.
Betanin attenuates paraquat-induced liver toxicity through a mitochondrial pathway.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-05-07)
Junyan Han, Zongju Zhang, Shaobin Yang, Jun Wang, Xuelian Yang, Dehong Tan
ABSTRACT
We attempted to determine whether betanin (from natural pigments) that has anti-oxidant properties would be protective against paraquat-induced liver injury in Sprague-Dawley rats. Paraquat was injected intraperitoneally into rats to induce liver toxicity. The rats were randomly divided into four groups: a control group, a paraquat group, and two groups that received betanin at doses of 25 and 100mg/kg/day three days before and two days after they were administered paraquat. We evaluated liver histopathology, serum liver enzymatic activities, oxidative stress, cytochrome P450 (CYP) 3A2 mRNA expression, and mitochondrial damage. The rats that were injected with paraquat incurred liver injury, evidenced by histological changes and elevated serum aspartate aminotransferase and alanine aminotransferase levels; paraquat also led to oxidative stress, an increase of cytochrome P450 3A2 mRNA expression, and mitochondrial damage, indicated by mitochondrial membrane swelling, reduced mitochondrial cytochrome C, and apoptosis-inducing factor protein levels. Pathological damage and all of the above mentioned markers were lesser in the animals treated with betanin than in those who received paraquat alone. Betanin had a protective effect against paraquat-induced liver damage in rats. The mechanism of the protection appears to be the inhibition of CYP 3A2 expression and protection of mitochondria.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C