  • Search Results
  • Critical role of bioanalytical strategies in investigation of clinical PK observations, a Phase I case study.
Critical role of bioanalytical strategies in investigation of clinical PK observations, a Phase I case study.
mAbs (2014-12-09)
Kun Peng, Keyang Xu, Luna Liu, Robert Hendricks, Reginald Delarosa, Rich Erickson, Nageshwar Budha, Maya Leabman, An Song, Surinder Kaur, Saloumeh K Fischer
ABSTRACT
RG7652 is a human immunoglobulin 1 (IgG1) monoclonal antibody (mAb) targeting proprotein convertase subtilisin/kexin type 9 (PCSK9) and is designed for the treatment of hypercholesterolemia. A target-binding enzyme-linked immunosorbent assay (ELISA) was developed to measure RG7652 levels in human serum in a Phase I study. Although target-binding assay formats are generally used to quantify free therapeutic, the actual therapeutic species being measured are affected by assay conditions, such as sample dilution and incubation time, and levels of soluble target in the samples. Therefore, in the presence of high concentrations of circulating target, the choice of reagents and assay conditions can have a significant effect on the observed pharmacokinetic (PK) profiles. Phase I RG7652 PK analysis using the ELISA data resulted in a nonlinear dose normalized exposure. An investigation was conducted to characterize the ELISA to determine whether the assay format and reagents may have contributed to the PK observation. In addition, to confirm the ELISA results, a second orthogonal method, liquid chromatography tandem mass spectrometry (LC-MS/MS) using a signature peptide as surrogate, was developed and implemented. A subset of PK samples, randomly selected from half of the subjects in the 6 single ascending dose (SAD) cohorts in the Phase I clinical study, was analyzed with the LC-MS/MS assay, and the data were found to be comparable to the ELISA data. This paper illustrates the importance of reagent characterization, as well as the benefits of using an orthogonal approach to eliminate bioanalytical contributions when encountering unexpected observations.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%