  • Search Results
  • Profiling of regioisomeric triacylglycerols in edible oils by supercritical fluid chromatography/tandem mass spectrometry.
Profiling of regioisomeric triacylglycerols in edible oils by supercritical fluid chromatography/tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-02-21)
Jae Won Lee, Toshiharu Nagai, Naohiro Gotoh, Eiichiro Fukusaki, Takeshi Bamba
ABSTRACT
In this study, supercritical fluid chromatography (SFC) coupled with triple quadrupole mass spectrometry was applied to the profiling of several regioisomeric triacylglycerols (TAGs). SFC conditions (column, flow rate, modifier) were optimized for the effective separation of TAGs. In the column test, a triacontyl (C30) silica gel reversed-phase column was selected to separate TAG regioisomers. Multiple reaction monitoring was used to selectively quantify each TAG. Then, the method was used to perform detailed characterization of a diverse array of TAGs in palm and canola oils. Seventy TAGs (C46:0-C60:2) of these oils were successfully analyzed as a result, and twenty isomeric TAG pairs were separated well. In particular, this method provided the fast and high resolution separation of six regioisomeric TAG pairs (PPLn/PLnP, PPL/PLP, PPO/POP, SPLn/SLnP, SPO/SOP, SSO/SOS-stearic acid (S, 18:0), oleic acid (O, 18:1), linoleic acid (L, 18:2), linolenic acid (Ln, 18:3), palmitic acid (P, 16:0)) in a short time (50min) as compared to high performance liquid chromatography. We were able to demonstrate the utility of this method for the analysis of regioisomeric TAGs in edible oils.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Supelco
Methanol, analytical standard
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
1-Oleoyl-rac-glycerol, ≥99%
Sigma-Aldrich
Glycerol, FCC, FG