  • Search Results
  • Tumor endothelial markers define novel subsets of cancer-specific circulating endothelial cells associated with antitumor efficacy.
Tumor endothelial markers define novel subsets of cancer-specific circulating endothelial cells associated with antitumor efficacy.
Cancer research (2014-03-15)
Reza Mehran, Monique Nilsson, Mehrdad Khajavi, Zhiqiang Du, Tina Cascone, Hua Kang Wu, Andrea Cortes, Li Xu, Amado Zurita, Robert Schier, Bernhard Riedel, Randa El-Zein, John V Heymach
ABSTRACT
Circulating endothelial cells (CEC) are derived from multiple sources, including bone marrow (circulating endothelial progenitors; CEP), and established vasculature (mature CEC). Although CECs have shown promise as a biomarker for patients with cancer, their utility has been limited, in part, by the lack of specificity for tumor vasculature and the different nonmalignant causes that can impact CEC. Tumor endothelial markers (TEM) are antigens enriched in tumor versus nonmalignant endothelia. We hypothesized that TEMs may be detectable on CEC and that these circulating TEM(+) endothelial cells (CTEC) may be a more specific marker for cancer and tumor response than standard CEC. We found that tumor-bearing mice had a relative increase in numbers of circulating CTEC, specifically with increased levels of TEM7 and TEM8 expression. Following treatment with various vascular-targeting agents, we observed a decrease in CTEC that correlated with the reductions in tumor growth. We extended these findings to human clinical samples and observed that CTECs were present in patients with esophageal cancer and non-small cell lung cancer (N = 40), and their levels decreased after surgical resection. These results demonstrate that CTECs are detectable in preclinical cancer models and patients with cancer. Furthermore, they suggest that CTECs offer a novel cancer-associated marker that may be useful as a blood-based surrogate for assessing the presence of tumor vasculature and antiangiogenic drug activity.
Sigma-Aldrich
Monoclonal Anti-PECAM1 antibody produced in mouse, clone 1D2-1A5, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-CD31 antibody produced in mouse, clone MEM-05, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, ACS reagent
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Supelco
Sucrose, analytical standard
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sucrose, Vetec, reagent grade, 99%
Sigma-Aldrich
Anti-PECAM1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard