  • Search Results
  • Simultaneous quantification of chrysophanol and physcion in rat plasma by ultra fast liquid chromatography-tandem mass spectrometry and application of the technique to comparative pharmacokinetic studies of Radix et Rhei Rhizoma extract alone and Dahuang Fuzi Decoction.
Simultaneous quantification of chrysophanol and physcion in rat plasma by ultra fast liquid chromatography-tandem mass spectrometry and application of the technique to comparative pharmacokinetic studies of Radix et Rhei Rhizoma extract alone and Dahuang Fuzi Decoction.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-01-20)
Xiao Liu, Huan Li, Li Wu, Jiangwa Xing, Yanhong Poh, Hao Cai, Bao Chang Cai
ABSTRACT
Most herbal medicines are prescribed in combination based on the theory of TCM to obtain synergistic effects or diminish the possible adverse reactions. Compatibility refers to the combination of two or more herbs based on the clinical settings and the properties of herbs. Chrysophanol and physcion are the main effective compounds in Radix et Rhizoma Rhei and Dahuang Fuzi Decoction which is the combination of Radix et Rhizoma Rhei, Radix Aconiti Lateralis Praeparata and Radix et Rhizoma Asari. However, chrysophanol and physcion are difficult to detect in vivo because of their low concentration and interference from endogenous compounds. The aim of this study is to develop a rapid, specific and sensitive ultra high performance liquid chromatography-triple quadrupole tandem mass method to simultaneously quantify chrysophanol and physcion in rat plasma, in order to better understand the pharmacokinetics and compatibility mechanism of Dahuang Fuzi Decoction for the first time. Multiple reaction monitoring (MRM) mode was applied for the quantitation at [M-H](-)m/z 253.0→m/z 225.1 for chrysophanol, [M-H](-) for m/z 283.1→m/z 240.0 physcion and [M-H](-)m/z 239.0→m/z 211.0 for IS. The analytes were separated on an Agilent Eclipse plus C18 column (100mm×2.1mm, 1.8μm) column within a total running time of 6.5min using a mixture of 3mM ammonium acetate in water and methanol (95:5, v/v) with a time program flow gradient according to the "plus gradient chromatography" theory. The inclusion of the ammonium acetate in the UFLC mobile phase dramatically improved the detection limit of the tested compounds and decreased the interference by matrix effects, which have been referred to as "LC-electrolyte effects". Finally, we demonstrated the application of a validated method in a comparative pharmacokinetic study of rats receiving an oral dose of Dahuang Fuzi Decoction or Radix et Rhei Rhizoma, the monarch drug in the prescription. Pharmacokinetic parameters showed significant difference between the two groups. The achieved comparative pharmacokinetic results were helpful to clarify the combination mechanism of Dahuang Fuzi Decoction.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Supelco
Ammonium acetate, suitable for mass spectrometry (MS), LiChropur, eluent additive for LC-MS
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)