  • Search Results
  • Enhancement on reactive oxygen species and COX-1 mRNA levels modulate the vascular relaxation induced by sodium nitroprusside in denuded mice aorta.
Enhancement on reactive oxygen species and COX-1 mRNA levels modulate the vascular relaxation induced by sodium nitroprusside in denuded mice aorta.
Fundamental & clinical pharmacology (2015-01-27)
Lucas M Kangussu, Vania C Olivon, Raquel D do N Arifa, Natália Araújo, Daniela Reis, Marieta T de A Assis, Frederico M Soriani, Daniele da G de Souza, Lusiane M Bendhack, Daniella Bonaventura
ABSTRACT
This study aimed to investigate the modulation of nitric oxide/reactive oxygen species in sodium nitroprusside relaxation in mice aorta. Sodium nitroprusside induced relaxation in endothelium-intact (e+) and endothelium-denuded (e-) aortas with greater potency in e+ than in e-. The nitric oxide synthase inhibitor did not alter the sodium nitroprusside relaxation in both e+ and e- aortas. However, the superoxide anion scavenger abolished the difference in sodium nitroprusside potency between e+ and e-. Sodium nitroprusside reduced dihydroethidium-derived fluorescent products in both groups; however, the difference between intact and denuded mice aorta remains. The glutathione levels and basal antioxidant activity of superoxide dismutase were reduced in e- aorta when compared with e+, and these values were not altered by sodium nitroprusside. Confirming these results, the levels of lipid peroxidation in e+ were significantly lower when compared to e-, and these values were not altered by sodium nitroprusside. The sodium nitroprusside potency in the presence of a nonselective COX inhibitor or the EP/DP prostaglandin receptor antagonist in endothelium denuded was similar to that in intact mice aorta. Based on these results, we performed the COX-1 and COX-2 mRNA level studies, and in denuded mice aorta, there was an upregulation in COX-1 mRNA levels. Taken together, our findings show that in the absence of endothelium, there is an enhancement of superoxide levels, leading to GSH consumption and higher levels of lipid peroxidation, showing an intense redox status. Furthermore, in denuded mice aorta, there was an upregulation of COX-1 mRNA expression, leading to vasoconstrictor prostanoids synthesis. The interaction of vasoconstrictor prostanoids with its receptors EP/DP negatively modulates the vascular relaxation induced by SNP in denuded mice aorta.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
Pyrogallol, ACS reagent, ≥99%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Pyrogallol, ≥98% (HPLC)
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Pyrogallol, ACS reagent
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
2-Nitrobenzoic acid, 95%, Contains 3- and 4-isomers
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
2-Nitrobenzoic acid, Vetec, reagent grade, 94%
Sigma-Aldrich
Sodium chloride, Vetec, reagent grade, 99%
Sigma-Aldrich
Pyrogallol, Vetec, reagent grade
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Pyrogallol, analytical standard
Sigma-Aldrich
Sodium chloride solution, 0.85%