  • Search Results
  • Salmonella enterica infection stimulates macrophages to hemophagocytose.
Salmonella enterica infection stimulates macrophages to hemophagocytose.
mBio (2014-12-11)
M Carolina Pilonieta, Sarah M Moreland, Christopher N English, Corrella S Detweiler
ABSTRACT
Hemophagocytes are cells of the monocyte lineage that have engulfed erythrocytes and leukocytes. Hemophagocytes frequently accumulate in patients with severe acute bacterial infections, such as those caused by Salmonella enterica, Brucella abortus, and Mycobacterium tuberculosis. The relationship between hemophagocytosis and infection is not well understood. In the murine liver, S. enterica serovar Typhimurium resides within hemophagocytic macrophages containing leukocytes. Here we show that S. Typhimurium also resides within hemophagocytes containing erythrocytes. In cell culture, S. Typhimurium benefits from residence within hemophagocytes by accessing iron, but why macrophages hemophagocytose is unknown. We show that treatment of macrophages with a cocktail of the proinflammatory cytokine interferon gamma (IFN-γ) and lipopolysaccharide (LPS) stimulates engulfment of nonsenescent erythrocytes. Exposure of resting or IFN-γ-treated macrophages to live, but not to heat-killed, S. Typhimurium cells also stimulates erythrocyte engulfment. Single-cell analyses show that S. Typhimurium-infected macrophages are more likely to erythrophagocytose and that infected macrophages engulf more erythrocytes than uninfected macrophages within the same culture well. In addition, macrophages containing erythrocytes harbor more bacteria. However, S. Typhimurium does not promote macrophage engulfment of polystyrene beads, suggesting a role for a ligand on the target cell. Finally, neither of the two S. Typhimurium type 3 secretion systems, T3SS1 or T3SS2, is fully required for hemophagocytosis. These results indicate that infection of macrophages with live S. Typhimurium cells stimulates hemophagocytosis. Macrophages are white blood cells (leukocytes) that engulf and destroy pathogens. Hemophagocytes, a subset of macrophages, are characteristic of severe acute infection in patients with, for instance, typhoid fever, brucellosis, tuberculosis, and leishmaniasis. Each of these diseases has the potential to become chronic. Hemophagocytes (blood-eating cells) engulf and degrade red and white blood cells for unknown reasons. The bacterial pathogen Salmonella acquires the essential nutrient iron from murine hemophagocytes. We report that Salmonella stimulates macrophages to engulf blood cells, indicating that cells of this bacterium actively promote the formation of a specialized cellular niche in which they can acquire nutrients, evade killing by the host immune system, and potentially transition to chronic infection.
Product Description
Supelco
Aphidicolin, analytical standard
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Diethyl azodicarboxylate solution, purum, ~40% in toluene (H-NMR)
Supelco
Sucrose, analytical standard
Sigma-Aldrich
L-Glutamine
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, Vetec, reagent grade, ≥99%
Sigma-Aldrich
Sucrose, Vetec, reagent grade, 99%