  • Search Results
  • The N terminus of type III secretion needle protein YscF from Yersinia pestis functions to modulate innate immune responses.
The N terminus of type III secretion needle protein YscF from Yersinia pestis functions to modulate innate immune responses.
Infection and immunity (2015-02-04)
Patrick Osei-Owusu, Danielle L Jessen Condry, Melody Toosky, William Roughead, David S Bradley, Matthew L Nilles
ABSTRACT
The type III secretion system is employed by many pathogens, including the genera Yersinia, Shigella, Pseudomonas, and Salmonella, to deliver effector proteins into eukaryotic cells. The injectisome needle is formed by the polymerization of a single protein, e.g., YscF (Yersinia pestis), PscF (Pseudomonas aeruginosa), PrgI (Salmonella enterica SPI-1), SsaG (Salmonella enterica SPI-2), or MxiH (Shigella flexneri). In this study, we demonstrated that the N termini of some needle proteins, particularly the N terminus of YscF from Yersinia pestis, influences host immune responses. The N termini of several needle proteins were truncated and tested for the ability to induce inflammatory responses in a human monocytic cell line (THP-1 cells). Truncated needle proteins induced proinflammatory cytokines to different magnitudes than the corresponding wild-type proteins, except SsaG. Notably, N-terminally truncated YscF induced significantly higher activation of NF-κB and/or AP-1 and higher induction of proinflammatory cytokines, suggesting that a function of the N terminus of YscF is interference with host sensing of YscF, consistent with Y. pestis pathogenesis. To directly test the ability of the N terminus of YscF to suppress cytokine induction, a YscF-SsaG chimera with 15 N-terminal amino acids from YscF added to SsaG was constructed. The chimeric YscF-SsaG induced lower levels of cytokines than wild-type SsaG. However, the addition of 15 random amino acids to SsaG had no effect on NF-κB/AP-1 activation. These results suggest that the N terminus of YscF can function to decrease cytokine induction, perhaps contributing to a favorable immune environment leading to survival of Y. pestis within the eukaryotic host.
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, Vetec, reagent grade, 99%
Sigma-Aldrich
L-Glutamine, Vetec, reagent grade, ≥99%
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E422, anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Glutamine, certified reference material, TraceCERT®
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
L-(+)-Arabinose, ≥99%