  • Search Results
  • Feasibility of haloperidol-anchored albumin nanoparticles loaded with doxorubicin as dry powder inhaler for pulmonary delivery.
Feasibility of haloperidol-anchored albumin nanoparticles loaded with doxorubicin as dry powder inhaler for pulmonary delivery.
Pharmaceutical development and technology (2013-11-14)
Jaleh Varshosaz, Farshid Hassanzadeh, Amin Mardani, Mahboubeh Rostami
ABSTRACT
Haloperidol (Hal) is a ligand that can target sigma 2 receptors over-expressed in non-small cell lung cancer. Hal targeted nanoparticles of bovine serum albumin (BSA) were prepared for pulmonary delivery of doxorubicin (DOX). The conjugation was confirmed by Fourier transform infrared spectroscopy (FTIR) and (1)H nuclear magnetic resonance ((1)H NMR) spectroscopic methods. Nanoparticles were prepared by desolvation method from BSA-Hal and were loaded with DOX. They were characterized for their morphology, particle size, zeta potential, drug loading and release efficiency. The optimized nanoparticles were spray-dried using trehalose, l-leucin and mannitol as dry powder inhaler (DPI) in different inlet temperatures between 80 and 120°C. The obtained nanocomposites were characterized for their aerodynamic diameter, specific surface area (cm(2)/g) and fine particle fraction (FPF) by a Cascade Impactor device. The optimized nanoparticles showed particle size of 218 nm, zeta potential of -25.4 mV, drug entrapment efficiency of 89% and release efficiency of 56% until 2 h. After spray drying of these nanoparticles, the best results were obtained from mannitol with an inlet temperature of 80°C which produced a mean aerodynamic diameter of 4.58 μm, FPF of 66% and specific surface area of 6302.99 cm(2)/g. The obtained results suggest that the designed DPI could be a suitable inhaler for targeted delivery of DOX in pulmonary delivery.
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
D-Mannitol, ≥98%
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
1,2-Dichloroethane, ACS reagent, ≥99.0%
Sigma-Aldrich
Succinic anhydride, ≥99% (GC)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
N,O-Bis(trimethylsilyl)acetamide, synthesis grade, ≥95%