  • Search Results
  • Apogossypolone inhibits the proliferation of LNCaP cells in vitro and in vivo.
Apogossypolone inhibits the proliferation of LNCaP cells in vitro and in vivo.
Molecular medicine reports (2014-07-26)
Xianqing Zhang, Xingbin Hu, Shijie Mu, Yonghua Zhan, Qunxing An, Zhixin Liu, Xiaofeng Huang
ABSTRACT
The aim of the present study was to investigate the anti‑tumor effect of apogossypolone (ApoG2) on human LNCaP cells in vitro and in vivo. Cell viability was evaluated using an MTT assay. Cell autophagy and apoptosis were detected by flow cytometry and using a terminal deoxynucleotidyl transferase dUTP nick end labeling assay, respectively. Morphological autophagy alterations were observed by transmission electron microscopy. The formation of acidic vesicular organelles was assessed by acridine orange staining and fluorescence microscopy. Quantitative polymerase chain reaction (qPCR) was conducted to detect the expression levels of apoptosis‑associated protein B‑cell lymphoma 2 (Bcl‑2) and Bak. The models of transplantation tumors in nude mice were established via subcutaneous injection of LNCaP cells. Growth of LNCaP cells was inhibited by ApoG2 treatment. Flow cytometry demonstrated that ApoG2 induced apoptosis in LNCaP cells. The Bcl‑2 expression was decreased while Bak expression was increased. In addition, activation of cysteine aspartate protease (caspase)‑3 and ‑8 was observed and 3‑methyladenine (3‑MA) enhanced apoptosis of LNCaP cells. Furthermore, nude mice treated with ApoG2 demonstrated a significant decrease in tumor volume and a significant increase in cell viability. Immunohistochemical analysis of tumor tissues demonstrated that ApoG2 enhanced caspase‑3, ‑8, LC‑3B and beclin‑1 expression and reduced the expression of Bcl‑2. ApoG2 was able to effectively suppress the growth of LNCaP cells through the induction of autophagy and apoptosis.
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O