  • Search Results
  • Liquid-phase microextraction based on two immiscible organic solvents followed by gas chromatography with mass spectrometry as an efficient method for the preconcentration and determination of cocaine, ketamine, and lidocaine in human urine samples.
Liquid-phase microextraction based on two immiscible organic solvents followed by gas chromatography with mass spectrometry as an efficient method for the preconcentration and determination of cocaine, ketamine, and lidocaine in human urine samples.
Journal of separation science (2014-06-26)
Yadollah Yamini, Shahram Seidi, Rouhollah Feizbakhsh, Tahmineh Baheri, Maryam Rezazadeh
ABSTRACT
A new type of liquid-phase microextraction based on two immiscible organic solvents was optimized and validated for the quantification of lidocaine, ketamine, and cocaine in human urine samples. A hollow-fiber based microextraction technique followed by gas chromatography coupled with mass spectrometry detection was used to reduce matrix interferences and improve limits of detection. The analytes were extracted from aqueous sample with pH 11.0, into a thin layer of organic solvent (n-dodecane) sustained in the pores of a hollow fiber, and then into a second organic acceptor (acetonitrile) located inside the lumen of the hollow fiber. With the application of optimized values, good linearity was obtained in the range of 1-500 μg/L for lidocaine and ketamine and 2-500 μg/L for cocaine with the determination coefficient values (r(2) ) >0.9943. The preconcentration factors and limits of detection (S/N > 3) were 250-350 and 0.01-0.05 μg/L, respectively. Intra and interassay precision values were <7.3 and 9.3%, respectively. The method was successfully applied for the determination and quantification of target analytes in human urine samples.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Dodecane, anhydrous, ≥99%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Dodecane, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
Dodecane, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Density Standard 749 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)