  • Search Results
  • Frozen adipose-derived mesenchymal stem cells maintain high capability to grow and differentiate.
Frozen adipose-derived mesenchymal stem cells maintain high capability to grow and differentiate.
Cryobiology (2014-07-19)
Greta Minonzio, Mattia Corazza, Luca Mariotta, Mauro Gola, Michele Zanzi, Eugenio Gandolfi, Domenico De Fazio, Gianni Soldati
ABSTRACT
In recent years, there has been a shift toward tissue-engineering strategies using stem cells for plastic and reconstructive surgical procedures. Therefore, it is important to develop safe and reproducible protocols for the extraction of adipose-derived stromal cells (ASCs) to allow cells to be stored in liquid nitrogen for future needs. The aspirated liposuction obtained from healthy donors were immediately processed after the suction using a protocol developed in our laboratory. The resulting stromal vascular fraction (SVF) was then characterized by the presence of adipose-derived stromal cells, at later stage frozen in liquid nitrogen. After that, cells were thawed and again characterized by adipose-derived stromal cells, cellular survival, differentiation ability and Colony Forming Unit-Fibroblast like colonies (CFU-F). Extraction and freezing of cells contained in the stromal vascular fraction demonstrate that thawed cells maintain the full capability to grow and differentiate in culture. The advent of adipose-derived stromal cells use in tissue engineering will assume a wide role in esthetic restoration in plastic surgery. It is thus important to develop clinically translatable protocols for the preparation and storage of adipose-derived stromal cells. Our results show that adipose-derived stromal cells in serum free can easily be frozen and stored in liquid nitrogen with retention of 85% of cell viability and 180,890 cell/g yield plus normal proliferative capacity and differentiation potential compared with fresh controls. These observations set the basis for adipose-derived stromal cells banking.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of Ph. Eur., BP, 35 wt. %, contains 10% methanol as stabilizer
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)