  • Search Results
  • A kinetic study of the main guaco metabolites using syrup formulation and the identification of an alternative route of coumarin metabolism in humans.
A kinetic study of the main guaco metabolites using syrup formulation and the identification of an alternative route of coumarin metabolism in humans.
PloS one (2015-03-11)
João Cleverson Gasparetto, Rosângela Gonçalves Peccinini, Thais Martins Guimarães de Francisco, Letícia Bonâncio Cerqueira, Francinete Ramos Campos, Roberto Pontarolo
ABSTRACT
For decades guaco species have been empirically used for the treatment of respiratory diseases. However, studies have shown that the toxic and therapeutic effects of the main guaco metabolites are dose-dependent, and none clinical study was done to evaluate the behavior of these substances in humans. In this work, a pilot study measuring the kinetic profile of the main guaco metabolites was performed leading to the knowledge of an alternative route of coumarin metabolism in humans. Initial screenings demonstrated that the administration of 60 mL of guaco syrup (single dose) did not provide sufficient levels of coumarin (COU), 7-hydroxycoumarin (7-HCOU), o-coumaric acid (OCA) and kaurenoic acid (KAU). The pharmacokinetic parameters were calculated by orally administering 60 mL of guaco syrup spiked with 1500 mg of COU. The kinetic study demonstrated that the plasmatic levels of 7-HCOU (considered the main metabolite of COU) were 10 times lower than the levels of COU, and the kinetic profile of 7-HCOU suggests sequential metabolism in the liver with low access of 7-HCOU to the systemic circulation. The study also demonstrated that OCA is one of the main bioavailable metabolites of COU. Therefore, the hydrolysis of the lactone ring forming a carboxylated compound is one of the possible routes of COU metabolism in humans. The half-lives of COU, 7-HCOU and OCA were approximately 4.0, 1.0 and 3.0 h, respectively and there was evidence that the recommended dosage of guaco syrup did not provide sufficient levels of COU, 7-HCOU or OCA to obtain a bronchodilation effect. Clinical studies are necessary to prove the efficacy and safety of products based on guaco.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
N-Ethylmaleimide, crystalline, ≥98% (HPLC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
tert-Butyl methyl ether, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Zirconyl chloride octahydrate, reagent grade, 98%