  • Search Results
  • Simultaneous determination of total nitrogen and total phosphorus in environmental waters using alkaline persulfate digestion and ion chromatography.
Simultaneous determination of total nitrogen and total phosphorus in environmental waters using alkaline persulfate digestion and ion chromatography.
Journal of chromatography. A (2014-12-03)
Brian M De Borba, Richard F Jack, Jeffrey S Rohrer, Joan Wirt, Dongmei Wang
ABSTRACT
An ion chromatography (IC) method was developed for the simultaneous determination of total nitrogen and total phosphorus after alkaline persulfate digestion. This study takes advantage of advances in construction of high-resolution, high-capacity anion-exchange columns that can better tolerate the matrices typically encountered when a determination of total nitrogen and total phosphorous is required. Here, we used an electrolytically generated hydroxide eluent combined with a high-capacity, hydroxide-selective, anion-exchange column for the determination of total nitrogen (as nitrate-N) and total phosphorus (as phosphate-P) in environmental samples by IC. This method yielded LODs for nitrate-N and phosphate-P of 1.0 and 1.3 μg/L, respectively. The LOQs determined for these analytes were 3.4 and 4.2 μg/L, respectively. Due to the dilution factor required and the blank nitrate-N concentration after the persulfate digestion, the quantification limits increased for nitrate-N and phosphate-P to 171 and 63 μg/L, respectively. The suitability of the method was evaluated by determining the nitrogen and phosphorus concentrations from known concentrations of organic-containing nitrogen and phosphorus compounds. In addition, environmental samples consisting of six different wastewaters and 48 reservoir samples were evaluated for total nitrogen and phosphorus. The recoveries of nitrogen and phosphorus from the organic-containing compounds ranged from 93.1 to 100.1% and 85.2 to 97.1%, respectively. In addition, good correlation between results obtained by the colorimetric method and IC was also observed. The linearity, accuracy, and evaluation of potential interferences for determining TN and TP will be discussed.
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium persulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Nicotinic acid, ≥98%
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Ammonium chloride, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O