  • Search Results
  • Coagulation assays and plasma fibrinogen concentrations in real-world patients with atrial fibrillation treated with dabigatran.
Coagulation assays and plasma fibrinogen concentrations in real-world patients with atrial fibrillation treated with dabigatran.
British journal of clinical pharmacology (2014-03-07)
Paul K L Chin, David M Patterson, Mei Zhang, Berit P Jensen, Daniel F B Wright, Murray L Barclay, Evan J Begg
ABSTRACT
In patients with atrial fibrillation prescribed dabigatran, the aim was to examine the correlation between plasma dabigatran concentrations and the three screening coagulation assays [international normalized ratio (INR), activated partial thromboplastin time (aPTT) and thrombin time (TT)] as well as the dilute thrombin time (dTT) and to examine the contribution of plasma fibrinogen concentrations to the variability in TT results. Plasma from patients with atrial fibrillation on dabigatran were analysed for clotting times and concentrations of fibrinogen and dabigatran. Correlation plots (and associated r(2) values) were generated using these data. The variability in TT results explained by fibrinogen concentrations was quantified using linear regression. Fifty-two patients (38-94 years old) contributed 120 samples, with plasma dabigatran concentrations ranging from 9 to 408 μg l(-1) . The r(2) values of INR, aPTT, TT and dTT against plasma dabigatran concentrations were 0.49, 0.54, 0.70 and 0.95, respectively. Plasma fibrinogen concentrations explained some of the residual variability in TT values after taking plasma dabigatran concentrations into account (r(2) = 0.12, P = 0.02). Of the screening coagulation assays, the TT correlated best with plasma dabigatran concentrations. Variability in fibrinogen concentrations accounts for some of the variability in the TT.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Supelco
Acetonitrile, analytical standard
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)