  • Search Results
  • Determination of zolpidem in human plasma by liquid chromatography-tandem mass spectrometry for clinical application.
Determination of zolpidem in human plasma by liquid chromatography-tandem mass spectrometry for clinical application.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-03-03)
Ji-Yeong Byeon, Hye-In Lee, Yun-Jeong Lee, Jung-Eun Lee, Se-Hyung Kim, Young-Hoon Kim, Han-Sung Na, Choon-Gon Jang, Seok-Yong Lee
ABSTRACT
Zolpidem (ZPD) is widely described for the short-term treatment of insomnia. We have developed and validated a simple and rapid liquid chromatography analytical method using tandem mass spectrometry (LC-MS/MS) for the quantification of ZPD in human plasma. Using dibucaine as an internal standard (IS), the analyte was extracted with methyl t-butyl ether (MTBE). Chromatographic separation of ZPD was performed on a reversed-phase Luna C18 column (50 mm × 2.0 mm i.d., 5 μm particles) with a mobile phase of 10mM ammonium formate buffer (pH 3.0)-methanol (15:85, v/v) at a flow rate of 250 μm/min. The total run-time was 2.5 min and the retention times of ZPD and IS were 0.66 and 0.74 min, respectively. The mass-to-charge transition monitored for quantification of ZPD and IS was 308.2→235.2 and 344.0→271.0, respectively. The lower limit of quantification (LLOQ) using 100 μL of human plasma was 0.05 ng/mL and the calibration curves were linear over a range of 0.05-200 ng/mL (r(2)>0.9964). The mean accuracy and precision for intra- and inter-run validation of ZPD were within acceptable limits. In the present LC-MS/MS method, we showed improved sensitivity for quantification of the ZPD in human plasma using lower volume of plasma compared with previously described analytical methods for ZPD. This validated method was successfully applied to a pharmacokinetic study in humans.
Product Description
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%