  • Search Results
  • Effect of resin infiltration on enamel surface properties and Streptococcus mutans adhesion to artificial enamel lesions.
Effect of resin infiltration on enamel surface properties and Streptococcus mutans adhesion to artificial enamel lesions.
Dental materials journal (2015-03-10)
Soley Arslan, Yahya Orcun Zorba, Mustafa Altay Atalay, Suat Özcan, Sezer Demirbuga, Kansad Pala, Duygu Percin, Fusun Ozer
ABSTRACT
The aim of this study was to evaluate and compare the effects of resin infiltration and sealant type on enamel surface properties and Streptococcus mutans adhesion to artificial enamel lesions. Artificial enamel lesions were produced on the surfaces of 120 enamel specimens, which were divided into two groups: Group A and Group B (n=60 per group). Each group was further divided into four subgroups (n=15 per subgroup) according to sealant type: Group I-Demineralized enamel (control); Group II-Enamel Pro Varnish; Group III-ExciTE F; and Group IV-Icon. In Group A, hardness and surface roughness were evaluated; in Group B, bacterial adhesion was evaluated. Icon application resulted in significantly lower surface roughness and higher hardness than the other subgroups in Group A. In Group B, Enamel Pro Varnish resulted in lowest bacterial adhesion, followed by Icon. This study showed that resin infiltration of enamel lesions could arrest lesion progress.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Silicic acid, BioReagent, suitable for column chromatography, 100-200 mesh (75 - 150 μm)
Sigma-Aldrich
Silicic acid, suitable for column chromatography, 60-200 mesh
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Silicic acid, BioReagent, suitable for column chromatography, 200-400 mesh (38 - 75 μm)
Sigma-Aldrich
Ethanol, absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC)
Sigma-Aldrich
Hydrogen chloride, puriss., ≥99.8%