The aim of the present work was to design a pH-modified solid dispersion (pH(M)-SD) that can improve the dissolution and bioavailability of poorly water-soluble weakly basic GT0918, a developing anti-prostate cancer drug. To select the appropriate acidifiers, a solubility test was carried out first. Solid dispersions (SDs) containing GT0918 and polyvinylpyrrolidone (PVP) were prepared using a solvent evaporation method and were characterized using dissolution studies in different media. The solid states of the SDs were investigated using scanning electron microscopy (SEM), powder X-ray diffraction (PXRD), differential scanning calorimetry (DSC) and Fourier transformed infrared spectroscopy (FTIR). The in vivo pharmacokinetics of the pH(M)-SDs tablets were also studied in beagle dogs compared to the conventional tablets. The optimized pH(M)-SD (GT0918/PVP/citric acid, 1:2:2 weight ratio) exhibited a significant improvement in the dissolution behavior compared to both the physical mixture and the binary SDs. Solid-state characterization revealed that the amorphous formation of GT0918 in the SDs and the strong H-bonding were only found in the pH(M)-SDs containing citric acid. Furthermore, the GT0918-loaded pH(M)-SD tablets showed a higher AUC and a lower tmax compared to the conventional tablets. Accordingly, the pH(M)-SD might be an efficient route for enhancing the dissolution and bioavailability of poorly water-soluble GT0918.