A novel inhaled multi-pronged attack against respiratory bacteria.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2015-01-24)
Sie Huey Lee, Jeanette Teo, Desmond Heng, Yanli Zhao, Wai Kiong Ng, Hak-Kim Chan, Li Teng Tan, Reginald B H Tan
ABSTRACT
Airway mucus hypersecretion is a common clinical feature of many severe respiratory diseases, and when complicated by a recalcitrant bacterial infection, the whole treatment regimen thereby becomes more challenging and protracted. The accumulation of thickened mucus secretions in the lower airways provides a nutrient-rich breeding ground for bacteria that promotes their growth and limits the ease of effective eradication. Unfortunately, no direct-inhaled dry powder formulation to treat these respiratory mucoid infections more effectively is available commercially. This work therefore seeks to develop a highly-efficacious ternary dry powder inhaler (DPI) formulation (ciprofloxacin hydrochloride (CIP), gatifloxacin hydrochloride (GAT) and ambroxol hydrochloride (AMB)) capable of delivering a novel multi-pronged attack (synergy, quorum quenching and mucociliary clearance) on Pseudomonas aeruginosa, a common respiratory bacteria found in mucoid infections. The powders were prepared via spray drying, evaluated on their aerosol performance via a multi-stage liquid impinger (MSLI) and tested for their efficacies in bacteria-spiked artificial sputum medium (ASM). The optimized particles were of respirable-size (d50 of ∼1.61±0.03μm) and slightly corrugated. When dispersed via an Aerolizer® inhaler at 60L/min, the powder showed concomitant in vitro deposition, minimal capsule, device and throat retention, and highly promising and uniform fine particle fractions (of the loaded dose) of ∼64-69%, which was a vast improvement over the singly-delivered actives. Favourably, when tested on bacteria-spiked ASM, the optimized ternary formulation (with AMB) was more effective at killing bacteria (i.e. faster rate of killing) than just the synergistic antibiotics alone (binary formulation; without AMB). In conclusion, a ternary antibiotic-(non-antibiotic) DPI formulation involving a unique multi-pronged attack mechanism was successfully pioneered and optimized for mucoid infections.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%
Sigma-Aldrich
D-Mannitol, ≥98%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥99% (titration)
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC