  • Search Results
  • Pharmacokinetic profiles of hydroxysafflor yellow A following intravenous administration of its pure preparations in healthy Chinese volunteers.
Pharmacokinetic profiles of hydroxysafflor yellow A following intravenous administration of its pure preparations in healthy Chinese volunteers.
Journal of ethnopharmacology (2015-01-13)
Chang-Yin Li, Jun-Gang Yin, Jun Zhang, Xiao-Xiao Wang, Mei-Juan Xu, Fang Liu, Jian-Dong Zou, Wen-Zheng Ju
ABSTRACT
Hydroxysafflor yellow A (HSYA), the major active marker compound isolated from Carthamus tinctorius L., has been demonstrated to possess various attractive pharmacological activities. However, there is a lack of information about the complete clinical pharmacokinetic profiles of HSYA following the administration of its pure preparations. The purpose of this study was to fully characterize the pharmacokinetic (PK) properties of HSYA in healthy Chinese volunteers following drip intravenous infusion of injectable powder of pure HSYA (IPPH), a new drug recently approved for the phase I clinical study by China Food and Drug Administration. 36 healthy subjects of either sex were recruited in this single-center, and open-label, single doses (25, 50, and 75 mg) and multiple doses (50 mg, once daily, 7 consecutive days) study. Plasma samples were analyzed with a validated LC-MS/MS method. Various PK parameters were estimated from the plasma concentration versus time data using non-compartmental methods. After single dose administration of IPPH, the values of AUC(0-t), AUC(0-∞) and C(max) for HSYA were statistically proportional over the dose range of 25-75 mg. After 7 repeated doses of 50 mg IPPH, both C(max) and AUC(0-∞) were significantly decreased, from 3207 to 2959 μg L(-1), and from 12,811 to 12,135 µg h L(-1) respectively, while t(1/2) was significantly prolonged from 3.912 to 4.414 h. The minimum plasma concentrations on day 5, 6 and 7 showed good stability with no significant difference. Both Cmax and AUC of HSYA in male volunteers were generally lower than that in females. IPPH was generally well tolerated in healthy volunteers by either single or multiple dosing. HSYA displayed moderately linear PK properties over the doses ranging from 25 to 75 mg of IPPH. Repeated administration of IPPH once daily could not lead to the in-vivo drug accumulation, but significantly affect PK behavior of HSYA. Gender difference should be considered for dosage recommendation in the clinic.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Ammonium acetate, suitable for mass spectrometry (MS), LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol, NMR reference standard