  • Search Results
  • Alteration of anti-inflammatory activity of Harpagophytum procumbens (devil's claw) extract after external metabolic activation with S9 mix.
Alteration of anti-inflammatory activity of Harpagophytum procumbens (devil's claw) extract after external metabolic activation with S9 mix.
The Journal of pharmacy and pharmacology (2014-09-02)
Katarina Hostanska, Joerg Melzer, Matthias Rostock, Andy Suter, Reinhard Saller
ABSTRACT
Extracts of the tubers of Harpagophytum procumbens (devil's claw, DC) inhibit different proinflammatory mediators important in the pathophysiology of osteoarthritis. Many plant-derived preparations interfere with cytochrome P450 liver enzymes, which influence their different biological activities. Therefore, the present study was designed to investigate the influence of an external metabolic activation of a DC extract on the cytotoxicity and the release of proinflammatory cytokines. A screening experiment with a panel of 12 inflammatory cytokines identified three as suitable for the study: tumour necrosis factor-α (TNF-α), interleukin (IL) IL-6 and IL-8. They were determined using enzyme-linked immunosorbent assays in lipopolysaccharide (LPS)-stimulated monocytic THP-1 cells, which were treated with rat liver S9 mix metabolically activated DC extract (DCm). For the cytotoxity experiments, a WST-1 assay was used. DC dose-dependently suppressed the release of TNF-α, IL-6 and IL-8 in LPS-stimulated monocytic THP-1 cells at non-cytotoxic concentrations (50-250 μg/ml). The metabolic activation of the DC extract by S9 mix did not alternate its cytotoxicity and did not diminish its inhibitory effect. This effect was improved in the case of TNF-α inhibition as reflected by their EC50 values of 116 ± 8.2 μg/ml and 49 ± 3.5 μg/ml for DC and DCm (P < 0.01). Cytokines inhibitory activity of DC was not affected after its external metabolic activation. However, the amount of harpagoside and caffeic acid derivates was decreased. Other components of the extract might have contributed to its anti-inflammatory effect.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Millipore
Acetonitrile solution, suitable for HPLC, acetonitrile:water 5:95% (v/v), 10 mM Ammoniumbicarbonate, pH 10,0
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube