  • Search Results
  • Target selective micelles for bombesin receptors incorporating Au(III)-dithiocarbamato complexes.
Target selective micelles for bombesin receptors incorporating Au(III)-dithiocarbamato complexes.
International journal of pharmaceutics (2014-07-12)
Paola Ringhieri, Roberta Iannitti, Chiara Nardon, Rosanna Palumbo, Dolores Fregona, Giancarlo Morelli, Antonella Accardo
ABSTRACT
Pure sterically stabilized micelles (SSM) of DSPE-PEG2000, and sterically stabilized mixed micelles (SSMM) containing PC or DOPC phospholipids (5, 10 or 20% mol/mol with respect to DSPE-PEG2000) are developed as delivery systems for the gold based cytotoxic drug Au(III)-dithiocarbamato complex AuL12. In particular, SSMM containing 5% of PC at 5mM of lipid concentration encapsulates 61.0 μg of AuL12 with a DL% of 1.13. The gold complex remains stable up to 72 h when incorporated in the aggregate, as indicated by UV-vis measurements. Incorporation in micelle composition of a low amount of the peptide derivative MonY-BN-AA1, containing a bombesin peptide analogue does not influence structural parameters of the micelles (diameter around 20 nm) neither the AuL12 loading parameters. Target selective properties of the peptide containing full aggregate on PC-3 cells overexpressing the GRP/bombesin receptors are observed by in vitro cytotoxic studies: a decrease of cell viability, ∼ 50%, is obtained in cells treated with AuL12-targeted micelles at 10 μM drug concentration for 48 h with respect to untargeted micelles.
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Sigma-Aldrich
1,2-Distearoyl-sn-glycero-3-phosphoethanolamine, ≥99%
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N