  • Search Results
  • Matrix solid-phase dispersion of polybrominated diphenyl ethers and their hydroxylated and methoxylated analogues in lettuce, carrot and soil.
Matrix solid-phase dispersion of polybrominated diphenyl ethers and their hydroxylated and methoxylated analogues in lettuce, carrot and soil.
Journal of chromatography. A (2014-08-19)
A Iparraguirre, R Rodil, J B Quintana, E Bizkarguenaga, A Prieto, O Zuloaga, R Cela, L A Fernández
ABSTRACT
In the present work, a novel analytical method for the simultaneous determination of ten polybrominated diphenyl ethers (PBDEs), eight methoxylated PBDEs (MeO-PBDEs) and seven hydroxylated PBDEs (OH-PBDEs) in soil, lettuce and carrot samples was developed. The procedure was based on matrix solid-phase dispersion (MSPD) followed by gas chromatography coupled to negative chemical ionization-mass spectrometry (GC-NCI-MS). Under optimum conditions, 0.5g of sample (freeze-dried in the case of lettuce and carrot samples) was dispersed with 0.5g of octadecyl-functionalized silica (C18) and 1.75g of acidified silica (10% H2SO4, w/w) was used as clean-up sorbent. A two-step fractionated elution was carried out. First, PBDEs and MeO-PBDEs were eluted in 75:25% (v/v) n-hexane/dichloromethane mixture and, then, the retained OH-PBDEs were eluted in pure dichloromethane. Both extracts were analyzed by GC-NCI-MS separately, in the case of OH-PBDEs after derivatization with N-methyl-N-(trimethylsilyl) trifluoroacetamide. The developed method was validated in terms of accuracy for soil, lettuce and carrot matrices, spiked at two fortification levels (5 and 25ngg(-1)). After correction with the corresponding surrogate, apparent recovery values (defined as the recovery obtained after correction with the corresponding surrogate) were in the 80-129% range. Precision (as relative standard deviation) in the 1-21% range and method detection limits (MDLs) in the 0.003 and 0.3ngg(-1) range for soil and in the 0.003-0.4ngg(-1) range (dry weight) for lettuce and carrot samples were obtained. For PBDEs the method was also validated with a standard reference material (SRM-2585) of house dust. Finally, the method was applied for the determination of target analytes in soil, lettuce and carrot.
Product Description
Methylene chloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 160 proof, Excise Tax-free, Permit for use required
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Ethanol, absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC)
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%